Assay Details
| Gene Symbol: BFSP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003571.3 | NP_003562.1 | 6 - 7 | 1335 | 112 | |
| GenBank mRNA | BC113518.1 | - | 6 - 7 | 1321 | 8322570 | 112 |
| BC113520.1 | - | 6 - 7 | 1321 | 8322572 | 112 | |
| BM667774.1 | - | 1 - 2 | 112 | |||
| U48224.1 | - | 6 - 7 | 1246 | 112 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03000720.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR03013020.hg.1 | TC03000720.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03013020.hg.1 | ENST00000302334.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03013020.hg.1 | NM_003571.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008856.hg.1 | ENST00000302334.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008856.hg.1 | ENST00000302334.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008856.hg.1 | ENST00000302334.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008856.hg.1 | ENST00000302334.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008856.hg.1 | ENST00000302334.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008856.hg.1 | ENST00000302334.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008856.hg.1 | ENST00000302334.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008856.hg.1 | ENST00000302334.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008856.hg.1 | ENST00000302334.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008856.hg.1 | ENST00000302334.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008856.hg.1 | ENST00000302334.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008856.hg.1 | ENST00000302334.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008856.hg.1 | ENST00000503047.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008856.hg.1 | ENST00000503047.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008856.hg.1 | ENST00000503047.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008856.hg.1 | ENST00000503047.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008856.hg.1 | ENST00000510039.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008856.hg.1 | ENST00000510039.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008856.hg.1 | ENST00000510039.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008856.hg.1 | ENST00000510039.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008856.hg.1 | ENST00000510039.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008856.hg.1 | ENST00000510039.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008856.hg.1 | ENST00000511140.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008856.hg.1 | ENST00000511140.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008856.hg.1 | ENST00000511434.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008856.hg.1 | ENST00000511434.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008856.hg.1 | ENST00000511434.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008856.hg.1 | ENST00000511434.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008856.hg.1 | ENST00000511434.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008856.hg.1 | ENST00000513441.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008856.hg.1 | ENST00000513441.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008856.hg.1 | NM_003571.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008856.hg.1 | NM_003571.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008856.hg.1 | NM_003571.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008856.hg.1 | NM_003571.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008856.hg.1 | NM_003571.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008856.hg.1 | NM_003571.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008856.hg.1 | NM_003571.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008856.hg.1 | NM_003571.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008856.hg.1 | NM_003571.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008856.hg.1 | NM_003571.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008856.hg.1 | NM_003571.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008856.hg.1 | NM_003571.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03013021.hg.1 | TC03000720.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03013021.hg.1 | ENST00000302334.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03013021.hg.1 | NM_003571.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03013022.hg.1 | TC03000720.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03013022.hg.1 | ENST00000302334.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03013022.hg.1 | NM_003571.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300163314.hg.1 | ENST00000302334.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300163314.hg.1 | ENST00000503047.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300163314.hg.1 | ENST00000510039.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300163314.hg.1 | NM_003571.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300163314.hg.1 | ENST00000302334.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300163314.hg.1 | ENST00000503047.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300163314.hg.1 | ENST00000510039.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300163314.hg.1 | NM_003571.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300163314.hg.1 | TC0300008856.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300008856.hg.1 | ENST00000302334.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008856.hg.1 | ENST00000302334.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008856.hg.1 | ENST00000302334.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008856.hg.1 | ENST00000302334.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008856.hg.1 | ENST00000302334.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008856.hg.1 | ENST00000302334.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008856.hg.1 | ENST00000302334.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008856.hg.1 | ENST00000302334.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008856.hg.1 | ENST00000302334.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008856.hg.1 | ENST00000302334.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008856.hg.1 | ENST00000302334.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008856.hg.1 | ENST00000302334.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008856.hg.1 | ENST00000503047.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008856.hg.1 | ENST00000503047.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008856.hg.1 | ENST00000503047.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008856.hg.1 | ENST00000503047.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008856.hg.1 | ENST00000510039.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008856.hg.1 | ENST00000510039.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008856.hg.1 | ENST00000510039.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008856.hg.1 | ENST00000510039.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008856.hg.1 | ENST00000510039.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008856.hg.1 | ENST00000510039.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008856.hg.1 | ENST00000511140.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008856.hg.1 | ENST00000511140.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008856.hg.1 | ENST00000511434.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008856.hg.1 | ENST00000511434.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008856.hg.1 | ENST00000511434.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008856.hg.1 | ENST00000511434.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008856.hg.1 | ENST00000511434.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008856.hg.1 | ENST00000513441.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008856.hg.1 | ENST00000513441.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008856.hg.1 | NM_003571.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008856.hg.1 | NM_003571.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008856.hg.1 | NM_003571.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008856.hg.1 | NM_003571.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008856.hg.1 | NM_003571.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008856.hg.1 | NM_003571.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008856.hg.1 | NM_003571.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008856.hg.1 | NM_003571.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008856.hg.1 | NM_003571.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008856.hg.1 | NM_003571.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008856.hg.1 | NM_003571.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008856.hg.1 | NM_003571.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300163312.hg.1 | ENST00000302334.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300163312.hg.1 | ENST00000503047.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300163312.hg.1 | ENST00000510039.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300163312.hg.1 | NM_003571.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300163312.hg.1 | ENST00000302334.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300163312.hg.1 | ENST00000503047.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300163312.hg.1 | ENST00000510039.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300163312.hg.1 | NM_003571.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300163312.hg.1 | TC0300008856.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300163309.hg.1 | ENST00000302334.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300163309.hg.1 | ENST00000503047.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300163309.hg.1 | ENST00000510039.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300163309.hg.1 | NM_003571.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300163309.hg.1 | ENST00000302334.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300163309.hg.1 | ENST00000503047.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300163309.hg.1 | ENST00000510039.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300163309.hg.1 | NM_003571.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300163309.hg.1 | TC0300008856.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 126010 | - | - |

BFSP2
Genomic Map