Assay Details
| Gene Symbol: DUSP12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007240.2 | NP_009171.1 | 1 - 2 | 378 | 62 | |
| GenBank mRNA | AF119226.1 | - | 1 - 2 | 355 | 62 | |
| BC006286.1 | - | 1 - 2 | 376 | 3958403 | 62 | |
| BT006633.1 | - | 1 - 2 | 344 | 62 | ||
| ES309317.1 | - | 1 - 2 | 394 | 62 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01001413.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
cellular protein modification process
dephosphorylation
positive regulation of glucokinase activity
peptidyl-tyrosine dephosphorylation
protein tyrosine phosphatase activity
protein binding
protein tyrosine/serine/threonine phosphatase activity
zinc ion binding
phosphatase activity
kinase binding
cellular protein modification process
dephosphorylation
positive regulation of glucokinase activity
peptidyl-tyrosine dephosphorylation
protein tyrosine phosphatase activity
protein binding
protein tyrosine/serine/threonine phosphatase activity
zinc ion binding
phosphatase activity
kinase binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01022107.hg.1 | TC01001413.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022107.hg.1 | ENST00000367943.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022107.hg.1 | ENST00000484291.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022107.hg.1 | NM_007240.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022108.hg.1 | TC01001413.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022108.hg.1 | ENST00000367943.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022108.hg.1 | ENST00000484291.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022108.hg.1 | NM_007240.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022111.hg.1 | TC01001413.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022111.hg.1 | ENST00000367943.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022111.hg.1 | ENST00000484291.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022111.hg.1 | NM_007240.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010368.hg.1 | ENST00000367943.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010368.hg.1 | ENST00000367943.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010368.hg.1 | ENST00000367943.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010368.hg.1 | ENST00000367943.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010368.hg.1 | ENST00000367943.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010368.hg.1 | ENST00000367943.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010368.hg.1 | ENST00000367943.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010368.hg.1 | ENST00000367943.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010368.hg.1 | ENST00000367943.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010368.hg.1 | ENST00000367943.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010368.hg.1 | ENST00000463365.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010368.hg.1 | ENST00000463365.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010368.hg.1 | ENST00000463365.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010368.hg.1 | ENST00000463365.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010368.hg.1 | ENST00000464004.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010368.hg.1 | ENST00000464004.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010368.hg.1 | ENST00000464004.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010368.hg.1 | ENST00000464004.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010368.hg.1 | ENST00000464004.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010368.hg.1 | ENST00000464004.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010368.hg.1 | ENST00000464004.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010368.hg.1 | ENST00000484291.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010368.hg.1 | ENST00000484291.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010368.hg.1 | ENST00000484291.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010368.hg.1 | ENST00000484291.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010368.hg.1 | ENST00000484291.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010368.hg.1 | ENST00000484291.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010368.hg.1 | ENST00000484291.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010368.hg.1 | ENST00000484291.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010368.hg.1 | ENST00000484291.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010368.hg.1 | ENST00000484291.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010368.hg.1 | ENST00000490591.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010368.hg.1 | ENST00000490591.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010368.hg.1 | ENST00000490591.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010368.hg.1 | NM_007240.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010368.hg.1 | NM_007240.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010368.hg.1 | NM_007240.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010368.hg.1 | NM_007240.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010368.hg.1 | NM_007240.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010368.hg.1 | NM_007240.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010368.hg.1 | NM_007240.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010368.hg.1 | NM_007240.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010368.hg.1 | NM_007240.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010368.hg.1 | NM_007240.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010368.hg.1 | ENST00000367943.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010368.hg.1 | ENST00000367943.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010368.hg.1 | ENST00000367943.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010368.hg.1 | ENST00000367943.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010368.hg.1 | ENST00000367943.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010368.hg.1 | ENST00000367943.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010368.hg.1 | ENST00000367943.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010368.hg.1 | ENST00000367943.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010368.hg.1 | ENST00000367943.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010368.hg.1 | ENST00000367943.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010368.hg.1 | ENST00000463365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010368.hg.1 | ENST00000463365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010368.hg.1 | ENST00000463365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010368.hg.1 | ENST00000463365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010368.hg.1 | ENST00000464004.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010368.hg.1 | ENST00000464004.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010368.hg.1 | ENST00000464004.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010368.hg.1 | ENST00000464004.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010368.hg.1 | ENST00000464004.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010368.hg.1 | ENST00000464004.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010368.hg.1 | ENST00000464004.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010368.hg.1 | ENST00000484291.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010368.hg.1 | ENST00000484291.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010368.hg.1 | ENST00000484291.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010368.hg.1 | ENST00000484291.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010368.hg.1 | ENST00000484291.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010368.hg.1 | ENST00000484291.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010368.hg.1 | ENST00000484291.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010368.hg.1 | ENST00000484291.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010368.hg.1 | ENST00000484291.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010368.hg.1 | ENST00000484291.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010368.hg.1 | ENST00000490591.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010368.hg.1 | ENST00000490591.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010368.hg.1 | ENST00000490591.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010368.hg.1 | NM_007240.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010368.hg.1 | NM_007240.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010368.hg.1 | NM_007240.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010368.hg.1 | NM_007240.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010368.hg.1 | NM_007240.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010368.hg.1 | NM_007240.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010368.hg.1 | NM_007240.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010368.hg.1 | NM_007240.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010368.hg.1 | NM_007240.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010368.hg.1 | NM_007240.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100176362.hg.1 | ENST00000367943.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176362.hg.1 | ENST00000464004.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176362.hg.1 | ENST00000484291.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176362.hg.1 | ENST00000490591.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176362.hg.1 | NM_007240.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100176362.hg.1 | ENST00000367943.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176362.hg.1 | ENST00000464004.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176362.hg.1 | ENST00000484291.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176362.hg.1 | ENST00000490591.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176362.hg.1 | NM_007240.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100176362.hg.1 | TC0100010368.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100176363.hg.1 | ENST00000367943.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176363.hg.1 | ENST00000464004.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176363.hg.1 | ENST00000484291.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176363.hg.1 | NM_007240.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100176363.hg.1 | ENST00000367943.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176363.hg.1 | ENST00000464004.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176363.hg.1 | ENST00000484291.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176363.hg.1 | NM_007240.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100176363.hg.1 | TC0100010368.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100176366.hg.1 | ENST00000367943.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176366.hg.1 | ENST00000464004.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176366.hg.1 | ENST00000484291.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176366.hg.1 | ENST00000490591.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176366.hg.1 | NM_007240.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100176366.hg.1 | ENST00000367943.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176366.hg.1 | ENST00000464004.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176366.hg.1 | ENST00000484291.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176366.hg.1 | ENST00000490591.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176366.hg.1 | NM_007240.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100176366.hg.1 | TC0100010368.hg.1 | Clariom D TC |

DUSP12
Genomic Map