Assay Details
Gene Symbol: NPTX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_002522.3 | NP_002513.2 | 1 - 2 | 598 | 86 | |
GenBank mRNA | AK127335.1 | - | 1 - 2 | 598 | 86 | |
BC089441.1 | - | 1 - 2 | 582 | 30555728 | 86 | |
U61849.1 | - | 1 - 2 | 572 | 86 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC17001937.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR17026390.hg.1 | ENST00000306773.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17026390.hg.1 | NM_002522.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17026390.hg.1 | TC17001937.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR17026391.hg.1 | ENST00000306773.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17026391.hg.1 | NM_002522.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17026391.hg.1 | TC17001937.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1700011984.hg.1 | ENST00000306773.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011984.hg.1 | ENST00000306773.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011984.hg.1 | ENST00000306773.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011984.hg.1 | ENST00000306773.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011984.hg.1 | ENST00000306773.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011984.hg.1 | ENST00000306773.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011984.hg.1 | ENST00000306773.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011984.hg.1 | ENST00000306773.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011984.hg.1 | ENST00000306773.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011984.hg.1 | ENST00000306773.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011984.hg.1 | ENST00000306773.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011984.hg.1 | ENST00000535681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011984.hg.1 | ENST00000535681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011984.hg.1 | ENST00000535681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011984.hg.1 | ENST00000535681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011984.hg.1 | ENST00000535681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011984.hg.1 | ENST00000535681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011984.hg.1 | ENST00000535681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011984.hg.1 | ENST00000571100.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011984.hg.1 | ENST00000571100.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011984.hg.1 | ENST00000571100.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011984.hg.1 | ENST00000571100.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011984.hg.1 | ENST00000575212.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011984.hg.1 | ENST00000575212.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011984.hg.1 | ENST00000575212.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011984.hg.1 | NM_002522.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011984.hg.1 | NM_002522.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011984.hg.1 | NM_002522.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011984.hg.1 | NM_002522.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011984.hg.1 | NM_002522.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011984.hg.1 | NM_002522.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011984.hg.1 | NM_002522.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011984.hg.1 | NM_002522.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011984.hg.1 | NM_002522.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011984.hg.1 | NM_002522.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011984.hg.1 | NM_002522.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011984.hg.1 | NM_002522.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700197897.hg.1 | ENST00000306773.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700197897.hg.1 | ENST00000571100.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700197897.hg.1 | ENST00000575212.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700197897.hg.1 | NM_002522.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700197897.hg.1 | ENST00000306773.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700197897.hg.1 | ENST00000571100.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700197897.hg.1 | ENST00000575212.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700197897.hg.1 | NM_002522.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700197897.hg.1 | TC1700011984.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1700197900.hg.1 | ENST00000306773.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700197900.hg.1 | NM_002522.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700197900.hg.1 | ENST00000306773.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700197900.hg.1 | NM_002522.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700197900.hg.1 | TC1700011984.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1700197899.hg.1 | ENST00000306773.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700197899.hg.1 | NM_002522.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700197899.hg.1 | ENST00000306773.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700197899.hg.1 | NM_002522.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700197899.hg.1 | TC1700011984.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1700011984.hg.1 | ENST00000306773.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011984.hg.1 | ENST00000306773.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011984.hg.1 | ENST00000306773.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011984.hg.1 | ENST00000306773.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011984.hg.1 | ENST00000306773.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011984.hg.1 | ENST00000306773.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011984.hg.1 | ENST00000306773.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011984.hg.1 | ENST00000306773.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011984.hg.1 | ENST00000306773.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011984.hg.1 | ENST00000306773.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011984.hg.1 | ENST00000306773.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011984.hg.1 | ENST00000535681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011984.hg.1 | ENST00000535681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011984.hg.1 | ENST00000535681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011984.hg.1 | ENST00000535681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011984.hg.1 | ENST00000535681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011984.hg.1 | ENST00000535681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011984.hg.1 | ENST00000535681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011984.hg.1 | ENST00000571100.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011984.hg.1 | ENST00000571100.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011984.hg.1 | ENST00000571100.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011984.hg.1 | ENST00000571100.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011984.hg.1 | ENST00000575212.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011984.hg.1 | ENST00000575212.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011984.hg.1 | ENST00000575212.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011984.hg.1 | NM_002522.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011984.hg.1 | NM_002522.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011984.hg.1 | NM_002522.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011984.hg.1 | NM_002522.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011984.hg.1 | NM_002522.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011984.hg.1 | NM_002522.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011984.hg.1 | NM_002522.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011984.hg.1 | NM_002522.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011984.hg.1 | NM_002522.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011984.hg.1 | NM_002522.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011984.hg.1 | NM_002522.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011984.hg.1 | NM_002522.3 | Refseq |
