Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011984.hg.1 |
ENST00000306773.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011984.hg.1 |
ENST00000306773.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011984.hg.1 |
ENST00000306773.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011984.hg.1 |
ENST00000306773.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011984.hg.1 |
ENST00000306773.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011984.hg.1 |
ENST00000306773.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011984.hg.1 |
ENST00000306773.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011984.hg.1 |
ENST00000306773.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011984.hg.1 |
ENST00000306773.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011984.hg.1 |
ENST00000306773.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011984.hg.1 |
ENST00000306773.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011984.hg.1 |
ENST00000535681.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011984.hg.1 |
ENST00000535681.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011984.hg.1 |
ENST00000535681.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011984.hg.1 |
ENST00000535681.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011984.hg.1 |
ENST00000535681.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011984.hg.1 |
ENST00000535681.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011984.hg.1 |
ENST00000535681.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011984.hg.1 |
ENST00000571100.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011984.hg.1 |
ENST00000571100.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011984.hg.1 |
ENST00000571100.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011984.hg.1 |
ENST00000571100.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011984.hg.1 |
ENST00000575212.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011984.hg.1 |
ENST00000575212.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011984.hg.1 |
ENST00000575212.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011984.hg.1 |
NM_002522.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011984.hg.1 |
NM_002522.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011984.hg.1 |
NM_002522.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011984.hg.1 |
NM_002522.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011984.hg.1 |
NM_002522.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011984.hg.1 |
NM_002522.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011984.hg.1 |
NM_002522.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011984.hg.1 |
NM_002522.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011984.hg.1 |
NM_002522.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011984.hg.1 |
NM_002522.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011984.hg.1 |
NM_002522.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011984.hg.1 |
NM_002522.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17026388.hg.1 |
ENST00000306773.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17026388.hg.1 |
ENST00000535681.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17026388.hg.1 |
NM_002522.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17026388.hg.1 |
TC17001937.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1700011984.hg.1 |
ENST00000306773.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011984.hg.1 |
ENST00000306773.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011984.hg.1 |
ENST00000306773.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011984.hg.1 |
ENST00000306773.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011984.hg.1 |
ENST00000306773.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011984.hg.1 |
ENST00000306773.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011984.hg.1 |
ENST00000306773.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011984.hg.1 |
ENST00000306773.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011984.hg.1 |
ENST00000306773.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011984.hg.1 |
ENST00000306773.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011984.hg.1 |
ENST00000306773.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011984.hg.1 |
ENST00000535681.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011984.hg.1 |
ENST00000535681.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011984.hg.1 |
ENST00000535681.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011984.hg.1 |
ENST00000535681.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011984.hg.1 |
ENST00000535681.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011984.hg.1 |
ENST00000535681.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011984.hg.1 |
ENST00000535681.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011984.hg.1 |
ENST00000571100.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011984.hg.1 |
ENST00000571100.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011984.hg.1 |
ENST00000571100.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011984.hg.1 |
ENST00000571100.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011984.hg.1 |
ENST00000575212.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011984.hg.1 |
ENST00000575212.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011984.hg.1 |
ENST00000575212.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011984.hg.1 |
NM_002522.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011984.hg.1 |
NM_002522.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011984.hg.1 |
NM_002522.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011984.hg.1 |
NM_002522.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011984.hg.1 |
NM_002522.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011984.hg.1 |
NM_002522.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011984.hg.1 |
NM_002522.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011984.hg.1 |
NM_002522.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011984.hg.1 |
NM_002522.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011984.hg.1 |
NM_002522.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011984.hg.1 |
NM_002522.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011984.hg.1 |
NM_002522.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700197893.hg.1 |
ENST00000306773.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700197893.hg.1 |
ENST00000535681.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700197893.hg.1 |
ENST00000571100.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700197893.hg.1 |
NM_002522.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700197893.hg.1 |
ENST00000306773.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700197893.hg.1 |
ENST00000535681.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700197893.hg.1 |
ENST00000571100.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700197893.hg.1 |
NM_002522.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700197893.hg.1 |
TC1700011984.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700197894.hg.1 |
ENST00000306773.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700197894.hg.1 |
ENST00000535681.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700197894.hg.1 |
ENST00000571100.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700197894.hg.1 |
ENST00000575212.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700197894.hg.1 |
NM_002522.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700197894.hg.1 |
ENST00000306773.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700197894.hg.1 |
ENST00000535681.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700197894.hg.1 |
ENST00000571100.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700197894.hg.1 |
ENST00000575212.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700197894.hg.1 |
NM_002522.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700197894.hg.1 |
TC1700011984.hg.1 |
Clariom D TC |