Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010825.hg.1 |
ENST00000375446.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010825.hg.1 |
ENST00000375446.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010825.hg.1 |
ENST00000375446.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010825.hg.1 |
ENST00000375446.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010825.hg.1 |
ENST00000375446.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010825.hg.1 |
ENST00000375446.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010825.hg.1 |
ENST00000375446.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010825.hg.1 |
ENST00000375446.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010825.hg.1 |
ENST00000461162.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010825.hg.1 |
ENST00000461162.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010825.hg.1 |
ENST00000461162.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010825.hg.1 |
ENST00000461162.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010825.hg.1 |
ENST00000461162.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010825.hg.1 |
ENST00000461162.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010825.hg.1 |
ENST00000470314.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010825.hg.1 |
ENST00000470314.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010825.hg.1 |
ENST00000470314.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010825.hg.1 |
ENST00000470314.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010825.hg.1 |
ENST00000470314.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010825.hg.1 |
ENST00000489274.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010825.hg.1 |
ENST00000489274.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010825.hg.1 |
ENST00000489274.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010825.hg.1 |
ENST00000489274.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010825.hg.1 |
ENST00000489274.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010825.hg.1 |
ENST00000490564.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010825.hg.1 |
ENST00000490564.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010825.hg.1 |
ENST00000490564.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010825.hg.1 |
ENST00000490564.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010825.hg.1 |
NM_004148.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010825.hg.1 |
NM_004148.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010825.hg.1 |
NM_004148.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010825.hg.1 |
NM_004148.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010825.hg.1 |
NM_004148.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010825.hg.1 |
NM_004148.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010825.hg.1 |
NM_004148.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010825.hg.1 |
NM_004148.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010825.hg.1 |
ENST00000375446.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010825.hg.1 |
ENST00000375446.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010825.hg.1 |
ENST00000375446.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010825.hg.1 |
ENST00000375446.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010825.hg.1 |
ENST00000375446.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010825.hg.1 |
ENST00000375446.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010825.hg.1 |
ENST00000375446.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010825.hg.1 |
ENST00000375446.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010825.hg.1 |
ENST00000461162.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010825.hg.1 |
ENST00000461162.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010825.hg.1 |
ENST00000461162.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010825.hg.1 |
ENST00000461162.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010825.hg.1 |
ENST00000461162.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010825.hg.1 |
ENST00000461162.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010825.hg.1 |
ENST00000470314.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010825.hg.1 |
ENST00000470314.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010825.hg.1 |
ENST00000470314.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010825.hg.1 |
ENST00000470314.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010825.hg.1 |
ENST00000470314.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010825.hg.1 |
ENST00000489274.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010825.hg.1 |
ENST00000489274.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010825.hg.1 |
ENST00000489274.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010825.hg.1 |
ENST00000489274.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010825.hg.1 |
ENST00000489274.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010825.hg.1 |
ENST00000490564.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010825.hg.1 |
ENST00000490564.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010825.hg.1 |
ENST00000490564.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010825.hg.1 |
ENST00000490564.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010825.hg.1 |
NM_004148.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010825.hg.1 |
NM_004148.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010825.hg.1 |
NM_004148.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010825.hg.1 |
NM_004148.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010825.hg.1 |
NM_004148.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010825.hg.1 |
NM_004148.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010825.hg.1 |
NM_004148.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010825.hg.1 |
NM_004148.3 |
Refseq |