Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR08009368.hg.1 |
TC08000694.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08009368.hg.1 |
NM_006438.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08009370.hg.1 |
TC08000694.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08009370.hg.1 |
ENST00000332843.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08009370.hg.1 |
ENST00000521788.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08009370.hg.1 |
NM_006438.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08009369.hg.1 |
TC08000694.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08009369.hg.1 |
ENST00000332843.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08009369.hg.1 |
NM_006438.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008658.hg.1 |
ENST00000332843.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008658.hg.1 |
ENST00000332843.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008658.hg.1 |
ENST00000332843.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008658.hg.1 |
ENST00000332843.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008658.hg.1 |
ENST00000332843.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008658.hg.1 |
ENST00000332843.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008658.hg.1 |
ENST00000332843.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008658.hg.1 |
ENST00000332843.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008658.hg.1 |
ENST00000332843.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008658.hg.1 |
ENST00000521788.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008658.hg.1 |
ENST00000521788.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008658.hg.1 |
ENST00000521788.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008658.hg.1 |
ENST00000521788.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008658.hg.1 |
ENST00000521788.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008658.hg.1 |
ENST00000521788.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008658.hg.1 |
ENST00000521788.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008658.hg.1 |
NM_006438.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008658.hg.1 |
NM_006438.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008658.hg.1 |
NM_006438.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008658.hg.1 |
NM_006438.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008658.hg.1 |
NM_006438.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008658.hg.1 |
NM_006438.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008658.hg.1 |
NM_006438.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008658.hg.1 |
NM_006438.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008658.hg.1 |
NM_006438.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008658.hg.1 |
NM_006438.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008658.hg.1 |
NM_006438.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800159345.hg.1 |
NM_006438.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800159345.hg.1 |
NM_006438.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800159345.hg.1 |
TC0800008658.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800159348.hg.1 |
ENST00000332843.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800159348.hg.1 |
ENST00000521788.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800159348.hg.1 |
NM_006438.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800159348.hg.1 |
ENST00000332843.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800159348.hg.1 |
ENST00000521788.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800159348.hg.1 |
NM_006438.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800159348.hg.1 |
TC0800008658.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800159346.hg.1 |
ENST00000332843.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800159346.hg.1 |
NM_006438.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800159346.hg.1 |
ENST00000332843.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800159346.hg.1 |
NM_006438.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800159346.hg.1 |
TC0800008658.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0800008658.hg.1 |
ENST00000332843.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008658.hg.1 |
ENST00000332843.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008658.hg.1 |
ENST00000332843.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008658.hg.1 |
ENST00000332843.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008658.hg.1 |
ENST00000332843.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008658.hg.1 |
ENST00000332843.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008658.hg.1 |
ENST00000332843.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008658.hg.1 |
ENST00000332843.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008658.hg.1 |
ENST00000332843.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008658.hg.1 |
ENST00000521788.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008658.hg.1 |
ENST00000521788.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008658.hg.1 |
ENST00000521788.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008658.hg.1 |
ENST00000521788.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008658.hg.1 |
ENST00000521788.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008658.hg.1 |
ENST00000521788.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008658.hg.1 |
ENST00000521788.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008658.hg.1 |
NM_006438.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008658.hg.1 |
NM_006438.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008658.hg.1 |
NM_006438.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008658.hg.1 |
NM_006438.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008658.hg.1 |
NM_006438.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008658.hg.1 |
NM_006438.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008658.hg.1 |
NM_006438.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008658.hg.1 |
NM_006438.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008658.hg.1 |
NM_006438.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008658.hg.1 |
NM_006438.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008658.hg.1 |
NM_006438.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800159347.hg.1 |
ENST00000332843.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800159347.hg.1 |
NM_006438.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800159347.hg.1 |
ENST00000332843.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800159347.hg.1 |
NM_006438.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800159347.hg.1 |
TC0800008658.hg.1 |
Clariom D TC |