| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14013487.hg.1 |
TC14002325.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14013487.hg.1 |
ENST00000247194.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14013487.hg.1 |
ENST00000487285.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14013487.hg.1 |
NM_144581.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010757.hg.1 |
ENST00000247194.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010757.hg.1 |
ENST00000247194.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010757.hg.1 |
ENST00000247194.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010757.hg.1 |
ENST00000247194.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010757.hg.1 |
ENST00000247194.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010757.hg.1 |
ENST00000247194.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010757.hg.1 |
ENST00000247194.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010757.hg.1 |
ENST00000247194.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010757.hg.1 |
ENST00000247194.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010757.hg.1 |
ENST00000247194.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010757.hg.1 |
ENST00000247194.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010757.hg.1 |
ENST00000463432.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010757.hg.1 |
ENST00000463432.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010757.hg.1 |
ENST00000463432.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010757.hg.1 |
ENST00000463432.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010757.hg.1 |
ENST00000478430.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010757.hg.1 |
ENST00000478430.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010757.hg.1 |
ENST00000478430.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010757.hg.1 |
ENST00000478430.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010757.hg.1 |
ENST00000478430.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010757.hg.1 |
ENST00000481608.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010757.hg.1 |
ENST00000481608.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010757.hg.1 |
ENST00000481608.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010757.hg.1 |
ENST00000481608.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010757.hg.1 |
ENST00000481608.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010757.hg.1 |
ENST00000487285.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010757.hg.1 |
ENST00000487285.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010757.hg.1 |
ENST00000487285.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010757.hg.1 |
ENST00000487285.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010757.hg.1 |
ENST00000487285.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010757.hg.1 |
ENST00000527981.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010757.hg.1 |
ENST00000527981.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010757.hg.1 |
ENST00000527981.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010757.hg.1 |
ENST00000527981.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010757.hg.1 |
ENST00000527981.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010757.hg.1 |
ENST00000527981.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010757.hg.1 |
ENST00000527981.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010757.hg.1 |
ENST00000532049.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010757.hg.1 |
ENST00000532049.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010757.hg.1 |
ENST00000532049.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010757.hg.1 |
ENST00000532049.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010757.hg.1 |
ENST00000543619.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010757.hg.1 |
ENST00000543619.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010757.hg.1 |
ENST00000543619.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010757.hg.1 |
ENST00000543619.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010757.hg.1 |
ENST00000543619.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010757.hg.1 |
NM_144581.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010757.hg.1 |
NM_144581.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010757.hg.1 |
NM_144581.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010757.hg.1 |
NM_144581.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010757.hg.1 |
NM_144581.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010757.hg.1 |
NM_144581.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010757.hg.1 |
NM_144581.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010757.hg.1 |
NM_144581.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010757.hg.1 |
NM_144581.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14013489.hg.1 |
TC14002325.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14013489.hg.1 |
ENST00000247194.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14013489.hg.1 |
ENST00000487285.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14013489.hg.1 |
NM_144581.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010757.hg.1 |
ENST00000247194.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010757.hg.1 |
ENST00000247194.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010757.hg.1 |
ENST00000247194.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010757.hg.1 |
ENST00000247194.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010757.hg.1 |
ENST00000247194.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010757.hg.1 |
ENST00000247194.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010757.hg.1 |
ENST00000247194.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010757.hg.1 |
ENST00000247194.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010757.hg.1 |
ENST00000247194.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010757.hg.1 |
ENST00000247194.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010757.hg.1 |
ENST00000247194.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010757.hg.1 |
ENST00000463432.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010757.hg.1 |
ENST00000463432.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010757.hg.1 |
ENST00000463432.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010757.hg.1 |
ENST00000463432.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010757.hg.1 |
ENST00000478430.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010757.hg.1 |
ENST00000478430.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010757.hg.1 |
ENST00000478430.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010757.hg.1 |
ENST00000478430.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010757.hg.1 |
ENST00000478430.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010757.hg.1 |
ENST00000481608.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010757.hg.1 |
ENST00000481608.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010757.hg.1 |
ENST00000481608.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010757.hg.1 |
ENST00000481608.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010757.hg.1 |
ENST00000481608.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010757.hg.1 |
ENST00000487285.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010757.hg.1 |
ENST00000487285.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010757.hg.1 |
ENST00000487285.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010757.hg.1 |
ENST00000487285.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010757.hg.1 |
ENST00000487285.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010757.hg.1 |
ENST00000527981.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010757.hg.1 |
ENST00000527981.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010757.hg.1 |
ENST00000527981.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010757.hg.1 |
ENST00000527981.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010757.hg.1 |
ENST00000527981.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010757.hg.1 |
ENST00000527981.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010757.hg.1 |
ENST00000527981.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010757.hg.1 |
ENST00000532049.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010757.hg.1 |
ENST00000532049.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010757.hg.1 |
ENST00000532049.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010757.hg.1 |
ENST00000532049.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010757.hg.1 |
ENST00000543619.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010757.hg.1 |
ENST00000543619.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010757.hg.1 |
ENST00000543619.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010757.hg.1 |
ENST00000543619.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010757.hg.1 |
ENST00000543619.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010757.hg.1 |
NM_144581.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010757.hg.1 |
NM_144581.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010757.hg.1 |
NM_144581.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010757.hg.1 |
NM_144581.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010757.hg.1 |
NM_144581.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010757.hg.1 |
NM_144581.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010757.hg.1 |
NM_144581.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010757.hg.1 |
NM_144581.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010757.hg.1 |
NM_144581.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400182583.hg.1 |
ENST00000247194.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400182583.hg.1 |
ENST00000481608.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400182583.hg.1 |
ENST00000487285.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400182583.hg.1 |
NM_144581.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400182583.hg.1 |
ENST00000247194.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400182583.hg.1 |
ENST00000481608.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400182583.hg.1 |
ENST00000487285.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400182583.hg.1 |
NM_144581.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400182583.hg.1 |
TC1400010757.hg.1 |
Clariom D TC |