Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02002181.hg.1 |
TC02000168.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02002181.hg.1 |
ENST00000264705.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02002181.hg.1 |
ENST00000403525.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02002181.hg.1 |
NM_004341.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200148323.hg.1 |
ENST00000264705.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200148323.hg.1 |
ENST00000403525.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200148323.hg.1 |
ENST00000428460.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200148323.hg.1 |
NM_001306079.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200148323.hg.1 |
NM_004341.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200148323.hg.1 |
ENST00000264705.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200148323.hg.1 |
ENST00000403525.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200148323.hg.1 |
ENST00000428460.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200148323.hg.1 |
NM_001306079.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200148323.hg.1 |
NM_004341.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200148323.hg.1 |
TC0200007059.hg.1 |
Clariom D TC |