Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01039472.hg.1 |
TC01002534.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01039472.hg.1 |
ENST00000289248.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01039472.hg.1 |
ENST00000372985.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01039472.hg.1 |
ENST00000372990.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01039472.hg.1 |
ENST00000538156.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01039472.hg.1 |
NM_017821.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013778.hg.1 |
ENST00000289248.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013778.hg.1 |
ENST00000289248.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013778.hg.1 |
ENST00000289248.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013778.hg.1 |
ENST00000289248.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013778.hg.1 |
ENST00000289248.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013778.hg.1 |
ENST00000289248.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013778.hg.1 |
ENST00000289248.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013778.hg.1 |
ENST00000289248.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013778.hg.1 |
ENST00000289248.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013778.hg.1 |
ENST00000289248.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013778.hg.1 |
ENST00000372990.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013778.hg.1 |
ENST00000372990.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013778.hg.1 |
ENST00000372990.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013778.hg.1 |
ENST00000372990.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013778.hg.1 |
ENST00000372990.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013778.hg.1 |
ENST00000372990.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013778.hg.1 |
ENST00000372990.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013778.hg.1 |
ENST00000372990.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013778.hg.1 |
ENST00000372990.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013778.hg.1 |
ENST00000372990.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013778.hg.1 |
ENST00000372990.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013778.hg.1 |
ENST00000372990.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013778.hg.1 |
ENST00000540558.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013778.hg.1 |
ENST00000540558.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013778.hg.1 |
ENST00000540558.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013778.hg.1 |
ENST00000540558.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013778.hg.1 |
ENST00000540558.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013778.hg.1 |
NM_001304746.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013778.hg.1 |
NM_001304746.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013778.hg.1 |
NM_001304746.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013778.hg.1 |
NM_001304746.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013778.hg.1 |
NM_001304746.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013778.hg.1 |
NM_001304746.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013778.hg.1 |
NM_001304746.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013778.hg.1 |
NM_001304746.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013778.hg.1 |
NM_001304746.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013778.hg.1 |
NM_001304746.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013778.hg.1 |
NM_001304746.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013778.hg.1 |
NM_001304746.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013778.hg.1 |
NM_017821.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013778.hg.1 |
NM_017821.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013778.hg.1 |
NM_017821.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013778.hg.1 |
NM_017821.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013778.hg.1 |
NM_017821.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013778.hg.1 |
NM_017821.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013778.hg.1 |
NM_017821.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013778.hg.1 |
NM_017821.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013778.hg.1 |
NM_017821.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013778.hg.1 |
NM_017821.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013778.hg.1 |
NM_017821.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013778.hg.1 |
NM_017821.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013778.hg.1 |
NM_017821.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013778.hg.1 |
ENST00000289248.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013778.hg.1 |
ENST00000289248.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013778.hg.1 |
ENST00000289248.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013778.hg.1 |
ENST00000289248.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013778.hg.1 |
ENST00000289248.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013778.hg.1 |
ENST00000289248.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013778.hg.1 |
ENST00000289248.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013778.hg.1 |
ENST00000289248.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013778.hg.1 |
ENST00000289248.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013778.hg.1 |
ENST00000289248.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013778.hg.1 |
ENST00000372990.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013778.hg.1 |
ENST00000372990.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013778.hg.1 |
ENST00000372990.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013778.hg.1 |
ENST00000372990.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013778.hg.1 |
ENST00000372990.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013778.hg.1 |
ENST00000372990.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013778.hg.1 |
ENST00000372990.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013778.hg.1 |
ENST00000372990.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013778.hg.1 |
ENST00000372990.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013778.hg.1 |
ENST00000372990.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013778.hg.1 |
ENST00000372990.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013778.hg.1 |
ENST00000372990.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013778.hg.1 |
ENST00000540558.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013778.hg.1 |
ENST00000540558.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013778.hg.1 |
ENST00000540558.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013778.hg.1 |
ENST00000540558.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013778.hg.1 |
ENST00000540558.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013778.hg.1 |
NM_001304746.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013778.hg.1 |
NM_001304746.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013778.hg.1 |
NM_001304746.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013778.hg.1 |
NM_001304746.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013778.hg.1 |
NM_001304746.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013778.hg.1 |
NM_001304746.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013778.hg.1 |
NM_001304746.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013778.hg.1 |
NM_001304746.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013778.hg.1 |
NM_001304746.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013778.hg.1 |
NM_001304746.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013778.hg.1 |
NM_001304746.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013778.hg.1 |
NM_001304746.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013778.hg.1 |
NM_017821.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013778.hg.1 |
NM_017821.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013778.hg.1 |
NM_017821.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013778.hg.1 |
NM_017821.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013778.hg.1 |
NM_017821.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013778.hg.1 |
NM_017821.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013778.hg.1 |
NM_017821.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013778.hg.1 |
NM_017821.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013778.hg.1 |
NM_017821.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013778.hg.1 |
NM_017821.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013778.hg.1 |
NM_017821.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013778.hg.1 |
NM_017821.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013778.hg.1 |
NM_017821.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100202980.hg.1 |
ENST00000289248.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100202980.hg.1 |
ENST00000372990.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100202980.hg.1 |
NM_001304746.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100202980.hg.1 |
NM_017821.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100202980.hg.1 |
ENST00000289248.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100202980.hg.1 |
ENST00000372990.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100202980.hg.1 |
NM_001304746.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100202980.hg.1 |
NM_017821.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100202980.hg.1 |
TC0100013778.hg.1 |
Clariom D TC |