Assay Details
| Gene Symbol: TEKT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144705.3 | NP_653306.1 | 1 - 2 | 646 | 86 | |
| XM_005263876.2 | XP_005263933.1 | 1 - 2 | 4312 | 86 | ||
| XM_011510670.1 | XP_011508972.1 | 1 - 2 | 4312 | 86 | ||
| GenBank mRNA | BC021716.2 | - | 1 - 2 | 592 | 4831157 | 86 |
| BI832036.1 | - | 1 - 2 | 637 | 86 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02000566.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR02008647.hg.1 | TC02000566.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02008647.hg.1 | ENST00000295201.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02008647.hg.1 | ENST00000427593.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02008647.hg.1 | NM_144705.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02008650.hg.1 | TC02000566.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02008650.hg.1 | ENST00000295201.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02008650.hg.1 | NM_144705.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00007042.hg.1 | ENST00000620700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00007042.hg.1 | ENST00000620700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00007042.hg.1 | ENST00000620700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00007042.hg.1 | ENST00000620700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00007042.hg.1 | ENST00000620700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00007042.hg.1 | ENST00000620700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008447.hg.1 | ENST00000295201.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008447.hg.1 | ENST00000295201.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008447.hg.1 | ENST00000295201.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008447.hg.1 | ENST00000295201.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008447.hg.1 | ENST00000295201.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008447.hg.1 | ENST00000295201.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008447.hg.1 | ENST00000295201.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008447.hg.1 | ENST00000295201.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008447.hg.1 | ENST00000295201.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008447.hg.1 | ENST00000468063.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008447.hg.1 | ENST00000468063.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008447.hg.1 | ENST00000468063.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008447.hg.1 | ENST00000468063.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008447.hg.1 | ENST00000468063.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008447.hg.1 | ENST00000468063.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008447.hg.1 | ENST00000468063.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008447.hg.1 | ENST00000468063.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008447.hg.1 | ENST00000468063.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008447.hg.1 | ENST00000468063.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008447.hg.1 | NM_001286559.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008447.hg.1 | NM_001286559.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008447.hg.1 | NM_001286559.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008447.hg.1 | NM_001286559.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008447.hg.1 | NM_001286559.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008447.hg.1 | NM_001286559.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008447.hg.1 | NM_001286559.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008447.hg.1 | NM_001286559.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008447.hg.1 | NM_001286559.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008447.hg.1 | NM_001286559.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008447.hg.1 | NM_144705.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008447.hg.1 | NM_144705.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008447.hg.1 | NM_144705.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008447.hg.1 | NM_144705.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008447.hg.1 | NM_144705.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008447.hg.1 | NM_144705.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008447.hg.1 | NM_144705.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008447.hg.1 | NM_144705.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008447.hg.1 | NM_144705.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02008652.hg.1 | TC02000566.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02008652.hg.1 | ENST00000295201.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02008652.hg.1 | NM_144705.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02008651.hg.1 | TC02000566.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02008651.hg.1 | ENST00000295201.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02008651.hg.1 | NM_144705.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200158453.hg.1 | ENST00000295201.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200158453.hg.1 | ENST00000468063.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200158453.hg.1 | NM_001286559.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200158453.hg.1 | NM_144705.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200158453.hg.1 | ENST00000295201.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200158453.hg.1 | ENST00000468063.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200158453.hg.1 | NM_001286559.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200158453.hg.1 | NM_144705.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200158453.hg.1 | TC0200008447.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200158455.hg.1 | ENST00000295201.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200158455.hg.1 | ENST00000468063.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200158455.hg.1 | NM_001286559.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200158455.hg.1 | NM_144705.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200158455.hg.1 | ENST00000295201.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200158455.hg.1 | ENST00000468063.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200158455.hg.1 | NM_001286559.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200158455.hg.1 | NM_144705.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200158455.hg.1 | TC0200008447.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200158457.hg.1 | ENST00000295201.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200158457.hg.1 | ENST00000468063.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200158457.hg.1 | NM_001286559.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200158457.hg.1 | NM_144705.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200158457.hg.1 | ENST00000295201.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200158457.hg.1 | ENST00000468063.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200158457.hg.1 | NM_001286559.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200158457.hg.1 | NM_144705.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200158457.hg.1 | TC0200008447.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0Y00007042.hg.1 | ENST00000620700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00007042.hg.1 | ENST00000620700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00007042.hg.1 | ENST00000620700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00007042.hg.1 | ENST00000620700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00007042.hg.1 | ENST00000620700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00007042.hg.1 | ENST00000620700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200158458.hg.1 | ENST00000295201.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200158458.hg.1 | ENST00000468063.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200158458.hg.1 | NM_001286559.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200158458.hg.1 | NM_144705.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200158458.hg.1 | ENST00000295201.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200158458.hg.1 | ENST00000468063.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200158458.hg.1 | NM_001286559.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200158458.hg.1 | NM_144705.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200158458.hg.1 | TC0200008447.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200008447.hg.1 | ENST00000295201.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008447.hg.1 | ENST00000295201.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008447.hg.1 | ENST00000295201.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008447.hg.1 | ENST00000295201.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008447.hg.1 | ENST00000295201.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008447.hg.1 | ENST00000295201.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008447.hg.1 | ENST00000295201.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008447.hg.1 | ENST00000295201.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008447.hg.1 | ENST00000295201.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008447.hg.1 | ENST00000468063.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008447.hg.1 | ENST00000468063.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008447.hg.1 | ENST00000468063.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008447.hg.1 | ENST00000468063.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008447.hg.1 | ENST00000468063.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008447.hg.1 | ENST00000468063.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008447.hg.1 | ENST00000468063.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008447.hg.1 | ENST00000468063.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008447.hg.1 | ENST00000468063.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008447.hg.1 | ENST00000468063.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008447.hg.1 | NM_001286559.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008447.hg.1 | NM_001286559.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008447.hg.1 | NM_001286559.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008447.hg.1 | NM_001286559.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008447.hg.1 | NM_001286559.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008447.hg.1 | NM_001286559.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008447.hg.1 | NM_001286559.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008447.hg.1 | NM_001286559.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008447.hg.1 | NM_001286559.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008447.hg.1 | NM_001286559.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008447.hg.1 | NM_144705.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008447.hg.1 | NM_144705.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008447.hg.1 | NM_144705.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008447.hg.1 | NM_144705.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008447.hg.1 | NM_144705.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008447.hg.1 | NM_144705.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008447.hg.1 | NM_144705.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008447.hg.1 | NM_144705.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008447.hg.1 | NM_144705.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200158454.hg.1 | ENST00000295201.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200158454.hg.1 | ENST00000468063.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200158454.hg.1 | NM_001286559.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200158454.hg.1 | NM_144705.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200158454.hg.1 | ENST00000295201.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200158454.hg.1 | ENST00000468063.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200158454.hg.1 | NM_001286559.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200158454.hg.1 | NM_144705.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200158454.hg.1 | TC0200008447.hg.1 | Clariom D TC |

TEKT4
Genomic Map