Assay Details
Gene Symbol: ALG6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013339.3 | NP_037471.2 | 4 - 5 | 559 | 95 | |
GenBank mRNA | AF063604.2 | - | 4 - 5 | 514 | 95 | |
AF102851.1 | - | 4 - 5 | 492 | 95 | ||
AK022700.1 | - | 4 - 5 | 497 | 95 | ||
BC001253.1 | - | 4 - 5 | 491 | 3452687 | 95 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01000719.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
protein N-linked glycosylation
dolichol-linked oligosaccharide biosynthetic process
oligosaccharide-lipid intermediate biosynthetic process
dolichyl-phosphate-glucose-glycolipid alpha-glucosyltransferase activity
dolichyl pyrophosphate Man9GlcNAc2 alpha-1,3-glucosyltransferase activity
glucosyltransferase activity
protein N-linked glycosylation
dolichol-linked oligosaccharide biosynthetic process
oligosaccharide-lipid intermediate biosynthetic process
dolichyl-phosphate-glucose-glycolipid alpha-glucosyltransferase activity
dolichyl pyrophosphate Man9GlcNAc2 alpha-1,3-glucosyltransferase activity
glucosyltransferase activity
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0100008584.hg.1 | ENST00000263440.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008584.hg.1 | ENST00000263440.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008584.hg.1 | ENST00000263440.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008584.hg.1 | ENST00000263440.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008584.hg.1 | ENST00000263440.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008584.hg.1 | ENST00000263440.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008584.hg.1 | ENST00000263440.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008584.hg.1 | ENST00000263440.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008584.hg.1 | ENST00000263440.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008584.hg.1 | ENST00000263440.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008584.hg.1 | ENST00000263440.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008584.hg.1 | ENST00000263440.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008584.hg.1 | ENST00000263440.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008584.hg.1 | ENST00000263440.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008584.hg.1 | ENST00000263440.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008584.hg.1 | ENST00000263440.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008584.hg.1 | ENST00000263440.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008584.hg.1 | ENST00000371108.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008584.hg.1 | ENST00000371108.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008584.hg.1 | ENST00000371108.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008584.hg.1 | ENST00000371108.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008584.hg.1 | ENST00000371108.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008584.hg.1 | ENST00000371108.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008584.hg.1 | ENST00000371108.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008584.hg.1 | ENST00000371108.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008584.hg.1 | ENST00000371108.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008584.hg.1 | ENST00000371108.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008584.hg.1 | ENST00000371108.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008584.hg.1 | ENST00000371108.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008584.hg.1 | ENST00000371108.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008584.hg.1 | ENST00000371108.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008584.hg.1 | ENST00000371108.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008584.hg.1 | ENST00000371108.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008584.hg.1 | ENST00000371108.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008584.hg.1 | ENST00000371108.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008584.hg.1 | ENST00000371108.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008584.hg.1 | ENST00000371108.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008584.hg.1 | ENST00000462390.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008584.hg.1 | ENST00000462390.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008584.hg.1 | ENST00000462390.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008584.hg.1 | ENST00000462390.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008584.hg.1 | ENST00000465969.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008584.hg.1 | ENST00000465969.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008584.hg.1 | ENST00000465969.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008584.hg.1 | ENST00000465969.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008584.hg.1 | ENST00000465969.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008584.hg.1 | ENST00000465969.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008584.hg.1 | ENST00000487136.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008584.hg.1 | ENST00000487136.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008584.hg.1 | ENST00000487136.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008584.hg.1 | ENST00000487136.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008584.hg.1 | ENST00000487136.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008584.hg.1 | ENST00000487136.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008584.hg.1 | ENST00000494765.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008584.hg.1 | ENST00000494765.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008584.hg.1 | ENST00000494765.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008584.hg.1 | ENST00000494765.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008584.hg.1 | ENST00000494765.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008584.hg.1 | ENST00000494765.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008584.hg.1 | ENST00000603108.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008584.hg.1 | ENST00000603108.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008584.hg.1 | ENST00000603108.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008584.hg.1 | ENST00000603108.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008584.hg.1 | ENST00000603108.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008584.hg.1 | ENST00000603108.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008584.hg.1 | ENST00000603108.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008584.hg.1 | ENST00000603108.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008584.hg.1 | ENST00000603108.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008584.hg.1 | ENST00000603108.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008584.hg.1 | ENST00000603108.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008584.hg.1 | ENST00000603108.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008584.hg.1 | ENST00000603108.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008584.hg.1 | ENST00000603108.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008584.hg.1 | ENST00000603108.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008584.hg.1 | ENST00000603108.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008584.hg.1 | NM_013339.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008584.hg.1 | NM_013339.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008584.hg.1 | NM_013339.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008584.hg.1 | NM_013339.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008584.hg.1 | NM_013339.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008584.hg.1 | NM_013339.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008584.hg.1 | NM_013339.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008584.hg.1 | NM_013339.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008584.hg.1 | NM_013339.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008584.hg.1 | NM_013339.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008584.hg.1 | NM_013339.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008584.hg.1 | NM_013339.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008584.hg.1 | NM_013339.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008584.hg.1 | NM_013339.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008584.hg.1 | NM_013339.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008584.hg.1 | NM_013339.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008584.hg.1 | NM_013339.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008584.hg.1 | NM_013339.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008584.hg.1 | NM_013339.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008584.hg.1 | NM_013339.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01011392.hg.1 | TC01000719.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01011392.hg.1 | ENST00000263440.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01011392.hg.1 | ENST00000371108.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01011392.hg.1 | NM_013339.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01011393.hg.1 | TC01000719.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01011393.hg.1 | ENST00000263440.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01011393.hg.1 | ENST00000371108.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01011393.hg.1 | NM_013339.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100160868.hg.1 | ENST00000263440.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100160868.hg.1 | ENST00000371108.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100160868.hg.1 | ENST00000603108.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100160868.hg.1 | NM_013339.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100160868.hg.1 | ENST00000263440.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100160868.hg.1 | ENST00000371108.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100160868.hg.1 | ENST00000603108.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100160868.hg.1 | NM_013339.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100160868.hg.1 | TC0100008584.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0100008584.hg.1 | ENST00000263440.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008584.hg.1 | ENST00000263440.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008584.hg.1 | ENST00000263440.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008584.hg.1 | ENST00000263440.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008584.hg.1 | ENST00000263440.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008584.hg.1 | ENST00000263440.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008584.hg.1 | ENST00000263440.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008584.hg.1 | ENST00000263440.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008584.hg.1 | ENST00000263440.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008584.hg.1 | ENST00000263440.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008584.hg.1 | ENST00000263440.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008584.hg.1 | ENST00000263440.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008584.hg.1 | ENST00000263440.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008584.hg.1 | ENST00000263440.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008584.hg.1 | ENST00000263440.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008584.hg.1 | ENST00000263440.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008584.hg.1 | ENST00000263440.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008584.hg.1 | ENST00000371108.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008584.hg.1 | ENST00000371108.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008584.hg.1 | ENST00000371108.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008584.hg.1 | ENST00000371108.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008584.hg.1 | ENST00000371108.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008584.hg.1 | ENST00000371108.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008584.hg.1 | ENST00000371108.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008584.hg.1 | ENST00000371108.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008584.hg.1 | ENST00000371108.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008584.hg.1 | ENST00000371108.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008584.hg.1 | ENST00000371108.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008584.hg.1 | ENST00000371108.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008584.hg.1 | ENST00000371108.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008584.hg.1 | ENST00000371108.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008584.hg.1 | ENST00000371108.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008584.hg.1 | ENST00000371108.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008584.hg.1 | ENST00000371108.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008584.hg.1 | ENST00000371108.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008584.hg.1 | ENST00000371108.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008584.hg.1 | ENST00000371108.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008584.hg.1 | ENST00000462390.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008584.hg.1 | ENST00000462390.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008584.hg.1 | ENST00000462390.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008584.hg.1 | ENST00000462390.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008584.hg.1 | ENST00000465969.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008584.hg.1 | ENST00000465969.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008584.hg.1 | ENST00000465969.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008584.hg.1 | ENST00000465969.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008584.hg.1 | ENST00000465969.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008584.hg.1 | ENST00000465969.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008584.hg.1 | ENST00000487136.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008584.hg.1 | ENST00000487136.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008584.hg.1 | ENST00000487136.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008584.hg.1 | ENST00000487136.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008584.hg.1 | ENST00000487136.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008584.hg.1 | ENST00000487136.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008584.hg.1 | ENST00000494765.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008584.hg.1 | ENST00000494765.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008584.hg.1 | ENST00000494765.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008584.hg.1 | ENST00000494765.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008584.hg.1 | ENST00000494765.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008584.hg.1 | ENST00000494765.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008584.hg.1 | ENST00000603108.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008584.hg.1 | ENST00000603108.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008584.hg.1 | ENST00000603108.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008584.hg.1 | ENST00000603108.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008584.hg.1 | ENST00000603108.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008584.hg.1 | ENST00000603108.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008584.hg.1 | ENST00000603108.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008584.hg.1 | ENST00000603108.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008584.hg.1 | ENST00000603108.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008584.hg.1 | ENST00000603108.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008584.hg.1 | ENST00000603108.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008584.hg.1 | ENST00000603108.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008584.hg.1 | ENST00000603108.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008584.hg.1 | ENST00000603108.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008584.hg.1 | ENST00000603108.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008584.hg.1 | ENST00000603108.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008584.hg.1 | NM_013339.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008584.hg.1 | NM_013339.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008584.hg.1 | NM_013339.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008584.hg.1 | NM_013339.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008584.hg.1 | NM_013339.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008584.hg.1 | NM_013339.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008584.hg.1 | NM_013339.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008584.hg.1 | NM_013339.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008584.hg.1 | NM_013339.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008584.hg.1 | NM_013339.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008584.hg.1 | NM_013339.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008584.hg.1 | NM_013339.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008584.hg.1 | NM_013339.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008584.hg.1 | NM_013339.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008584.hg.1 | NM_013339.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008584.hg.1 | NM_013339.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008584.hg.1 | NM_013339.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008584.hg.1 | NM_013339.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008584.hg.1 | NM_013339.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008584.hg.1 | NM_013339.3 | Refseq |
