Assay Details
Gene Symbol: ZSCAN21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145914.2 | NP_666019.1 | 1 - 2 | 45 | 75 | |
XM_017012585.1 | XP_016868074.1 | 1 - 2 | 69 | 75 | ||
GenBank mRNA | AK292267.1 | - | 1 - 2 | 65 | 75 | |
AK313884.1 | - | 1 - 2 | 65 | 75 | ||
BC047309.1 | - | 1 - 2 | 38 | 5111890 | 75 | |
BI764964.1 | - | 1 - 2 | 45 | 75 | ||
CR749301.1 | - | 1 - 2 | 40 | 75 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC07000608.hg
Additional Information:
This assay is in the 5'UTR region of the AK292267.1,BC047309.1,CR749301.1,NM_145914.2,XM_017012585.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
positive regulation of transcription from RNA polymerase II promoter
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
protein binding
metal ion binding
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
positive regulation of transcription from RNA polymerase II promoter
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
protein binding
metal ion binding
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR07009064.hg.1 | TC07000608.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR07009064.hg.1 | ENST00000292450.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07009064.hg.1 | ENST00000379635.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07009064.hg.1 | ENST00000456748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07009064.hg.1 | ENST00000477297.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07009064.hg.1 | ENST00000543588.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07009064.hg.1 | NM_145914.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | ENST00000292450.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | ENST00000292450.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | ENST00000292450.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | ENST00000292450.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | ENST00000292450.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | ENST00000292450.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | ENST00000292450.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | ENST00000292450.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | ENST00000292450.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | ENST00000292450.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | ENST00000292450.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | ENST00000292450.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | ENST00000438937.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | ENST00000438937.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | ENST00000438937.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | ENST00000438937.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | ENST00000438937.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | ENST00000438937.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | ENST00000438937.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | ENST00000456748.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | ENST00000456748.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | ENST00000456748.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | ENST00000456748.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | ENST00000456748.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | ENST00000456748.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | ENST00000456748.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | ENST00000456748.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | ENST00000456748.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | ENST00000456748.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | ENST00000456748.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | ENST00000477297.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | ENST00000477297.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | ENST00000477297.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | ENST00000477297.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | ENST00000477297.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | ENST00000477297.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | ENST00000477297.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | ENST00000543588.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | ENST00000543588.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | ENST00000543588.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | ENST00000543588.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | ENST00000543588.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | ENST00000543588.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | ENST00000543588.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | ENST00000543588.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | ENST00000543588.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | ENST00000543588.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | ENST00000543588.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | NM_145914.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | NM_145914.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | NM_145914.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | NM_145914.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | NM_145914.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | NM_145914.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | NM_145914.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | NM_145914.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | NM_145914.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | NM_145914.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | NM_145914.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700008519.hg.1 | NM_145914.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR07009060.hg.1 | TC07000608.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR07009060.hg.1 | ENST00000292450.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07009060.hg.1 | ENST00000456748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07009060.hg.1 | ENST00000543588.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07009060.hg.1 | NM_145914.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR07009063.hg.1 | TC07000608.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR07009063.hg.1 | ENST00000292450.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07009063.hg.1 | ENST00000379635.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07009063.hg.1 | ENST00000456748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07009063.hg.1 | ENST00000477297.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07009063.hg.1 | NM_145914.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700159698.hg.1 | ENST00000292450.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700159698.hg.1 | ENST00000438937.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700159698.hg.1 | ENST00000456748.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700159698.hg.1 | ENST00000543588.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700159698.hg.1 | NM_145914.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700159698.hg.1 | ENST00000292450.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700159698.hg.1 | ENST00000438937.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700159698.hg.1 | ENST00000456748.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700159698.hg.1 | ENST00000543588.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700159698.hg.1 | NM_145914.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700159698.hg.1 | TC0700008519.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0700159702.hg.1 | ENST00000292450.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700159702.hg.1 | ENST00000438937.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700159702.hg.1 | ENST00000456748.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700159702.hg.1 | ENST00000477297.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700159702.hg.1 | ENST00000543588.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700159702.hg.1 | NM_145914.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700159702.hg.1 | ENST00000292450.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700159702.hg.1 | ENST00000438937.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700159702.hg.1 | ENST00000456748.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700159702.hg.1 | ENST00000477297.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700159702.hg.1 | ENST00000543588.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700159702.hg.1 | NM_145914.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700159702.hg.1 | TC0700008519.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | ENST00000292450.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | ENST00000292450.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | ENST00000292450.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | ENST00000292450.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | ENST00000292450.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | ENST00000292450.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | ENST00000292450.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | ENST00000292450.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | ENST00000292450.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | ENST00000292450.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | ENST00000292450.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | ENST00000292450.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | ENST00000438937.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | ENST00000438937.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | ENST00000438937.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | ENST00000438937.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | ENST00000438937.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | ENST00000438937.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | ENST00000438937.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | ENST00000456748.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | ENST00000456748.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | ENST00000456748.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | ENST00000456748.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | ENST00000456748.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | ENST00000456748.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | ENST00000456748.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | ENST00000456748.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | ENST00000456748.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | ENST00000456748.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | ENST00000456748.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | ENST00000477297.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | ENST00000477297.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | ENST00000477297.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | ENST00000477297.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | ENST00000477297.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | ENST00000477297.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | ENST00000477297.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | ENST00000543588.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | ENST00000543588.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | ENST00000543588.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | ENST00000543588.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | ENST00000543588.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | ENST00000543588.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | ENST00000543588.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | ENST00000543588.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | ENST00000543588.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | ENST00000543588.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | ENST00000543588.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | NM_145914.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | NM_145914.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | NM_145914.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | NM_145914.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | NM_145914.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | NM_145914.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | NM_145914.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | NM_145914.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | NM_145914.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | NM_145914.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | NM_145914.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0700008519.hg.1 | NM_145914.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700159701.hg.1 | ENST00000292450.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700159701.hg.1 | ENST00000438937.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700159701.hg.1 | ENST00000456748.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700159701.hg.1 | ENST00000477297.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700159701.hg.1 | ENST00000543588.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700159701.hg.1 | NM_145914.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700159701.hg.1 | ENST00000292450.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700159701.hg.1 | ENST00000438937.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700159701.hg.1 | ENST00000456748.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700159701.hg.1 | ENST00000477297.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700159701.hg.1 | ENST00000543588.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700159701.hg.1 | NM_145914.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700159701.hg.1 | TC0700008519.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0700159700.hg.1 | ENST00000292450.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700159700.hg.1 | ENST00000438937.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700159700.hg.1 | ENST00000456748.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700159700.hg.1 | ENST00000477297.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700159700.hg.1 | ENST00000543588.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700159700.hg.1 | NM_145914.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700159700.hg.1 | ENST00000292450.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700159700.hg.1 | ENST00000438937.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700159700.hg.1 | ENST00000456748.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700159700.hg.1 | ENST00000477297.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700159700.hg.1 | ENST00000543588.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700159700.hg.1 | NM_145914.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700159700.hg.1 | TC0700008519.hg.1 | Clariom D TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 184519 | - | - |
