Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR07009066.hg.1 |
TC07000608.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07009066.hg.1 |
ENST00000292450.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07009066.hg.1 |
ENST00000379635.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07009066.hg.1 |
ENST00000456748.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07009066.hg.1 |
ENST00000477297.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07009066.hg.1 |
ENST00000543588.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07009066.hg.1 |
NM_145914.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07009065.hg.1 |
TC07000608.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07009065.hg.1 |
ENST00000292450.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07009065.hg.1 |
ENST00000456748.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07009065.hg.1 |
ENST00000477297.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07009065.hg.1 |
ENST00000543588.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07009065.hg.1 |
NM_145914.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07009067.hg.1 |
TC07000608.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07009067.hg.1 |
ENST00000292450.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07009067.hg.1 |
ENST00000379635.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07009067.hg.1 |
ENST00000456748.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07009067.hg.1 |
ENST00000477297.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07009067.hg.1 |
ENST00000543588.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07009067.hg.1 |
NM_145914.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008519.hg.1 |
ENST00000292450.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008519.hg.1 |
ENST00000292450.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008519.hg.1 |
ENST00000292450.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008519.hg.1 |
ENST00000292450.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008519.hg.1 |
ENST00000292450.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008519.hg.1 |
ENST00000292450.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008519.hg.1 |
ENST00000292450.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008519.hg.1 |
ENST00000292450.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008519.hg.1 |
ENST00000292450.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008519.hg.1 |
ENST00000292450.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008519.hg.1 |
ENST00000292450.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008519.hg.1 |
ENST00000292450.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008519.hg.1 |
ENST00000438937.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008519.hg.1 |
ENST00000438937.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008519.hg.1 |
ENST00000438937.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008519.hg.1 |
ENST00000438937.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008519.hg.1 |
ENST00000438937.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008519.hg.1 |
ENST00000438937.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008519.hg.1 |
ENST00000438937.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008519.hg.1 |
ENST00000456748.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008519.hg.1 |
ENST00000456748.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008519.hg.1 |
ENST00000456748.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008519.hg.1 |
ENST00000456748.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008519.hg.1 |
ENST00000456748.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008519.hg.1 |
ENST00000456748.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008519.hg.1 |
ENST00000456748.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008519.hg.1 |
ENST00000456748.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008519.hg.1 |
ENST00000456748.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008519.hg.1 |
ENST00000456748.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008519.hg.1 |
ENST00000456748.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008519.hg.1 |
ENST00000477297.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008519.hg.1 |
ENST00000477297.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008519.hg.1 |
ENST00000477297.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008519.hg.1 |
ENST00000477297.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008519.hg.1 |
ENST00000477297.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008519.hg.1 |
ENST00000477297.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008519.hg.1 |
ENST00000477297.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008519.hg.1 |
ENST00000543588.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008519.hg.1 |
ENST00000543588.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008519.hg.1 |
ENST00000543588.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008519.hg.1 |
ENST00000543588.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008519.hg.1 |
ENST00000543588.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008519.hg.1 |
ENST00000543588.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008519.hg.1 |
ENST00000543588.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008519.hg.1 |
ENST00000543588.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008519.hg.1 |
ENST00000543588.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008519.hg.1 |
ENST00000543588.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008519.hg.1 |
ENST00000543588.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008519.hg.1 |
NM_145914.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008519.hg.1 |
NM_145914.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008519.hg.1 |
NM_145914.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008519.hg.1 |
NM_145914.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008519.hg.1 |
NM_145914.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008519.hg.1 |
NM_145914.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008519.hg.1 |
NM_145914.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008519.hg.1 |
NM_145914.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008519.hg.1 |
NM_145914.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008519.hg.1 |
NM_145914.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008519.hg.1 |
NM_145914.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008519.hg.1 |
NM_145914.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700159703.hg.1 |
ENST00000292450.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700159703.hg.1 |
ENST00000438937.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700159703.hg.1 |
ENST00000456748.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700159703.hg.1 |
ENST00000477297.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700159703.hg.1 |
NM_145914.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700159703.hg.1 |
ENST00000292450.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700159703.hg.1 |
ENST00000438937.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700159703.hg.1 |
ENST00000456748.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700159703.hg.1 |
ENST00000477297.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700159703.hg.1 |
NM_145914.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700159703.hg.1 |
TC0700008519.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700159705.hg.1 |
ENST00000292450.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700159705.hg.1 |
ENST00000438937.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700159705.hg.1 |
ENST00000456748.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700159705.hg.1 |
ENST00000477297.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700159705.hg.1 |
ENST00000543588.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700159705.hg.1 |
NM_145914.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700159705.hg.1 |
ENST00000292450.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700159705.hg.1 |
ENST00000438937.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700159705.hg.1 |
ENST00000456748.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700159705.hg.1 |
ENST00000477297.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700159705.hg.1 |
ENST00000543588.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700159705.hg.1 |
NM_145914.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700159705.hg.1 |
TC0700008519.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0700008519.hg.1 |
ENST00000292450.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008519.hg.1 |
ENST00000292450.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008519.hg.1 |
ENST00000292450.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008519.hg.1 |
ENST00000292450.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008519.hg.1 |
ENST00000292450.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008519.hg.1 |
ENST00000292450.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008519.hg.1 |
ENST00000292450.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008519.hg.1 |
ENST00000292450.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008519.hg.1 |
ENST00000292450.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008519.hg.1 |
ENST00000292450.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008519.hg.1 |
ENST00000292450.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008519.hg.1 |
ENST00000292450.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008519.hg.1 |
ENST00000438937.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008519.hg.1 |
ENST00000438937.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008519.hg.1 |
ENST00000438937.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008519.hg.1 |
ENST00000438937.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008519.hg.1 |
ENST00000438937.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008519.hg.1 |
ENST00000438937.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008519.hg.1 |
ENST00000438937.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008519.hg.1 |
ENST00000456748.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008519.hg.1 |
ENST00000456748.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008519.hg.1 |
ENST00000456748.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008519.hg.1 |
ENST00000456748.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008519.hg.1 |
ENST00000456748.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008519.hg.1 |
ENST00000456748.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008519.hg.1 |
ENST00000456748.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008519.hg.1 |
ENST00000456748.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008519.hg.1 |
ENST00000456748.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008519.hg.1 |
ENST00000456748.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008519.hg.1 |
ENST00000456748.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008519.hg.1 |
ENST00000477297.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008519.hg.1 |
ENST00000477297.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008519.hg.1 |
ENST00000477297.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008519.hg.1 |
ENST00000477297.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008519.hg.1 |
ENST00000477297.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008519.hg.1 |
ENST00000477297.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008519.hg.1 |
ENST00000477297.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008519.hg.1 |
ENST00000543588.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008519.hg.1 |
ENST00000543588.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008519.hg.1 |
ENST00000543588.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008519.hg.1 |
ENST00000543588.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008519.hg.1 |
ENST00000543588.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008519.hg.1 |
ENST00000543588.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008519.hg.1 |
ENST00000543588.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008519.hg.1 |
ENST00000543588.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008519.hg.1 |
ENST00000543588.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008519.hg.1 |
ENST00000543588.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008519.hg.1 |
ENST00000543588.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008519.hg.1 |
NM_145914.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008519.hg.1 |
NM_145914.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008519.hg.1 |
NM_145914.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008519.hg.1 |
NM_145914.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008519.hg.1 |
NM_145914.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008519.hg.1 |
NM_145914.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008519.hg.1 |
NM_145914.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008519.hg.1 |
NM_145914.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008519.hg.1 |
NM_145914.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008519.hg.1 |
NM_145914.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008519.hg.1 |
NM_145914.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008519.hg.1 |
NM_145914.2 |
Refseq |