Assay Details
Gene Symbol: EPHA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_005233.5 | NP_005224.2 | 10 - 11 | 2113 | 139 | |
XM_005264715.2 | XP_005264772.1 | 10 - 11 | 2035 | 139 | ||
XM_005264716.2 | XP_005264773.1 | 10 - 11 | 2108 | 139 | ||
GenBank mRNA | AF213459.1 | - | 10 - 11 | 2113 | 139 | |
AK291411.1 | - | 10 - 11 | 1946 | 139 | ||
BC063282.1 | - | 10 - 11 | 2017 | 30347891 | 139 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC03000468.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
cell adhesion
regulation of epithelial to mesenchymal transition
positive regulation of neuron projection development
cell migration
peptidyl-tyrosine phosphorylation
regulation of actin cytoskeleton organization
regulation of GTPase activity
ephrin receptor signaling pathway
regulation of focal adhesion assembly
regulation of microtubule cytoskeleton organization
cellular response to retinoic acid
fasciculation of sensory neuron axon
fasciculation of motor neuron axon
GPI-linked ephrin receptor activity
protein binding
ATP binding
cell adhesion
regulation of epithelial to mesenchymal transition
positive regulation of neuron projection development
cell migration
peptidyl-tyrosine phosphorylation
regulation of actin cytoskeleton organization
regulation of GTPase activity
ephrin receptor signaling pathway
regulation of focal adhesion assembly
regulation of microtubule cytoskeleton organization
cellular response to retinoic acid
fasciculation of sensory neuron axon
fasciculation of motor neuron axon
GPI-linked ephrin receptor activity
protein binding
ATP binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0300008029.hg.1 | ENST00000336596.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008029.hg.1 | ENST00000336596.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008029.hg.1 | ENST00000336596.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008029.hg.1 | ENST00000336596.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008029.hg.1 | ENST00000336596.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008029.hg.1 | ENST00000336596.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008029.hg.1 | ENST00000336596.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008029.hg.1 | ENST00000336596.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008029.hg.1 | ENST00000336596.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008029.hg.1 | ENST00000336596.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008029.hg.1 | ENST00000336596.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008029.hg.1 | ENST00000336596.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008029.hg.1 | ENST00000336596.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008029.hg.1 | ENST00000336596.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008029.hg.1 | ENST00000336596.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008029.hg.1 | ENST00000336596.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008029.hg.1 | ENST00000336596.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008029.hg.1 | ENST00000336596.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008029.hg.1 | ENST00000336596.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008029.hg.1 | ENST00000336596.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008029.hg.1 | ENST00000336596.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008029.hg.1 | ENST00000336596.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008029.hg.1 | ENST00000336596.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008029.hg.1 | ENST00000452448.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008029.hg.1 | ENST00000452448.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008029.hg.1 | ENST00000452448.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008029.hg.1 | ENST00000452448.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008029.hg.1 | ENST00000452448.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008029.hg.1 | ENST00000452448.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008029.hg.1 | ENST00000452448.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008029.hg.1 | ENST00000452448.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008029.hg.1 | ENST00000452448.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008029.hg.1 | ENST00000452448.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008029.hg.1 | ENST00000494014.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008029.hg.1 | ENST00000494014.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008029.hg.1 | ENST00000494014.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008029.hg.1 | ENST00000494014.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008029.hg.1 | ENST00000494014.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008029.hg.1 | ENST00000494014.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008029.hg.1 | ENST00000494014.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008029.hg.1 | ENST00000494014.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008029.hg.1 | ENST00000494014.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008029.hg.1 | ENST00000494014.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008029.hg.1 | ENST00000494014.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008029.hg.1 | ENST00000494014.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008029.hg.1 | ENST00000494014.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008029.hg.1 | ENST00000494014.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008029.hg.1 | ENST00000494014.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008029.hg.1 | ENST00000494014.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008029.hg.1 | ENST00000494014.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008029.hg.1 | ENST00000494014.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008029.hg.1 | ENST00000494014.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008029.hg.1 | NM_005233.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008029.hg.1 | NM_005233.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008029.hg.1 | NM_005233.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008029.hg.1 | NM_005233.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008029.hg.1 | NM_005233.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008029.hg.1 | NM_005233.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008029.hg.1 | NM_005233.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008029.hg.1 | NM_005233.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008029.hg.1 | NM_005233.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008029.hg.1 | NM_005233.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008029.hg.1 | NM_005233.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008029.hg.1 | NM_005233.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008029.hg.1 | NM_005233.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008029.hg.1 | NM_005233.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008029.hg.1 | NM_005233.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008029.hg.1 | NM_005233.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008029.hg.1 | NM_005233.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008029.hg.1 | NM_005233.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008029.hg.1 | NM_005233.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008029.hg.1 | NM_005233.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008029.hg.1 | NM_005233.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008029.hg.1 | NM_005233.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008029.hg.1 | NM_005233.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008029.hg.1 | NM_182644.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008029.hg.1 | NM_182644.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008029.hg.1 | NM_182644.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008029.hg.1 | NM_182644.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008029.hg.1 | NM_182644.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008029.hg.1 | NM_182644.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008029.hg.1 | NM_182644.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008029.hg.1 | NM_182644.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008029.hg.1 | NM_182644.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008029.hg.1 | NM_182644.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008029.hg.1 | NM_182644.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008029.hg.1 | NM_182644.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR03009084.hg.1 | TC03000468.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR03009084.hg.1 | ENST00000336596.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03009084.hg.1 | ENST00000494014.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03009084.hg.1 | NM_005233.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008029.hg.1 | ENST00000336596.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008029.hg.1 | ENST00000336596.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008029.hg.1 | ENST00000336596.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008029.hg.1 | ENST00000336596.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008029.hg.1 | ENST00000336596.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008029.hg.1 | ENST00000336596.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008029.hg.1 | ENST00000336596.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008029.hg.1 | ENST00000336596.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008029.hg.1 | ENST00000336596.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008029.hg.1 | ENST00000336596.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008029.hg.1 | ENST00000336596.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008029.hg.1 | ENST00000336596.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008029.hg.1 | ENST00000336596.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008029.hg.1 | ENST00000336596.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008029.hg.1 | ENST00000336596.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008029.hg.1 | ENST00000336596.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008029.hg.1 | ENST00000336596.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008029.hg.1 | ENST00000336596.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008029.hg.1 | ENST00000336596.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008029.hg.1 | ENST00000336596.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008029.hg.1 | ENST00000336596.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008029.hg.1 | ENST00000336596.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008029.hg.1 | ENST00000336596.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008029.hg.1 | ENST00000452448.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008029.hg.1 | ENST00000452448.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008029.hg.1 | ENST00000452448.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008029.hg.1 | ENST00000452448.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008029.hg.1 | ENST00000452448.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008029.hg.1 | ENST00000452448.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008029.hg.1 | ENST00000452448.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008029.hg.1 | ENST00000452448.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008029.hg.1 | ENST00000452448.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008029.hg.1 | ENST00000452448.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008029.hg.1 | ENST00000494014.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008029.hg.1 | ENST00000494014.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008029.hg.1 | ENST00000494014.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008029.hg.1 | ENST00000494014.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008029.hg.1 | ENST00000494014.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008029.hg.1 | ENST00000494014.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008029.hg.1 | ENST00000494014.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008029.hg.1 | ENST00000494014.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008029.hg.1 | ENST00000494014.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008029.hg.1 | ENST00000494014.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008029.hg.1 | ENST00000494014.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008029.hg.1 | ENST00000494014.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008029.hg.1 | ENST00000494014.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008029.hg.1 | ENST00000494014.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008029.hg.1 | ENST00000494014.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008029.hg.1 | ENST00000494014.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008029.hg.1 | ENST00000494014.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008029.hg.1 | ENST00000494014.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008029.hg.1 | ENST00000494014.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008029.hg.1 | NM_005233.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008029.hg.1 | NM_005233.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008029.hg.1 | NM_005233.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008029.hg.1 | NM_005233.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008029.hg.1 | NM_005233.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008029.hg.1 | NM_005233.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008029.hg.1 | NM_005233.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008029.hg.1 | NM_005233.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008029.hg.1 | NM_005233.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008029.hg.1 | NM_005233.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008029.hg.1 | NM_005233.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008029.hg.1 | NM_005233.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008029.hg.1 | NM_005233.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008029.hg.1 | NM_005233.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008029.hg.1 | NM_005233.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008029.hg.1 | NM_005233.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008029.hg.1 | NM_005233.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008029.hg.1 | NM_005233.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008029.hg.1 | NM_005233.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008029.hg.1 | NM_005233.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008029.hg.1 | NM_005233.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008029.hg.1 | NM_005233.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008029.hg.1 | NM_005233.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008029.hg.1 | NM_182644.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008029.hg.1 | NM_182644.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008029.hg.1 | NM_182644.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008029.hg.1 | NM_182644.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008029.hg.1 | NM_182644.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008029.hg.1 | NM_182644.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008029.hg.1 | NM_182644.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008029.hg.1 | NM_182644.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008029.hg.1 | NM_182644.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008029.hg.1 | NM_182644.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008029.hg.1 | NM_182644.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008029.hg.1 | NM_182644.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300157307.hg.1 | ENST00000336596.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300157307.hg.1 | ENST00000494014.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300157307.hg.1 | NM_005233.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300157307.hg.1 | ENST00000336596.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300157307.hg.1 | ENST00000494014.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300157307.hg.1 | NM_005233.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300157307.hg.1 | TC0300008029.hg.1 | Clariom D TC |
