Assay Details
Gene Symbol: TONSL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013432.4 | NP_038460.4 | 3 - 4 | 284 | 62 | |
XM_011517050.2 | XP_011515352.1 | 3 - 4 | 343 | 62 | ||
GenBank mRNA | BC008782.2 | - | 2 - 3 | 256 | 3628374 | 62 |
CN366457.1 | - | 3 - 4 | 308 | 62 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC08001751.hg
Additional Information:
This assay is in the 5'UTR region of the BC008782.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR08023288.hg.1 | TC08001751.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR08023288.hg.1 | ENST00000409379.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08023288.hg.1 | ENST00000422691.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08023288.hg.1 | NM_013432.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000613741.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000613741.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000613741.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000613741.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000613741.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000613741.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000613741.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000613741.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000613741.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000613741.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000613741.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000613741.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000613741.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000613741.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | ENST00000613741.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | NM_013432.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | NM_013432.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | NM_013432.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | NM_013432.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | NM_013432.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | NM_013432.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | NM_013432.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | NM_013432.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | NM_013432.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | NM_013432.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | NM_013432.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | NM_013432.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | NM_013432.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | NM_013432.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | NM_013432.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | NM_013432.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | NM_013432.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | NM_013432.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | NM_013432.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | NM_013432.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | NM_013432.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | NM_013432.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | NM_013432.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | NM_013432.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | NM_013432.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | NM_013432.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | NM_013432.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012214.hg.1 | NM_013432.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000409379.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000497613.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000613741.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000613741.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000613741.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000613741.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000613741.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000613741.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000613741.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000613741.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000613741.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000613741.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000613741.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000613741.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000613741.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000613741.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | ENST00000613741.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | NM_013432.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | NM_013432.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | NM_013432.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | NM_013432.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | NM_013432.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | NM_013432.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | NM_013432.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | NM_013432.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | NM_013432.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | NM_013432.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | NM_013432.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | NM_013432.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | NM_013432.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | NM_013432.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | NM_013432.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | NM_013432.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | NM_013432.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | NM_013432.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | NM_013432.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | NM_013432.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | NM_013432.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | NM_013432.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | NM_013432.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | NM_013432.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | NM_013432.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | NM_013432.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | NM_013432.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012214.hg.1 | NM_013432.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800183817.hg.1 | ENST00000409379.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800183817.hg.1 | ENST00000613741.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800183817.hg.1 | NM_013432.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800183817.hg.1 | ENST00000409379.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800183817.hg.1 | ENST00000613741.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800183817.hg.1 | NM_013432.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800183817.hg.1 | TC0800012214.hg.1 | Clariom D TC |
