| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000219281.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000219281.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000219281.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000219281.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000219281.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000219281.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000219281.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000219281.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000219281.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000219281.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000219281.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000219281.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000219281.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000219281.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000219281.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000219281.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000219281.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000423271.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000423271.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000423271.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000423271.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000423271.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000423271.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000423271.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000423271.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000423271.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000423271.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000423271.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000539737.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000539737.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000539737.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000539737.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000539737.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000539737.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000539737.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000539737.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000539737.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000539737.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000539737.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000539737.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000539737.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000539737.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000539737.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000561568.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000561568.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000561568.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000561568.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000561568.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000561568.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000561568.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000561568.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000561568.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000561743.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000561743.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000561743.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000561743.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000561743.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000561743.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000561743.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000561743.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000561743.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000561743.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000561743.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000562534.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000562534.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000562534.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000562534.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000562534.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000562534.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000562534.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000563149.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000563149.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000563149.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000563149.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000563149.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000563149.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000563149.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000563149.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000563207.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000563207.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000563207.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000563207.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000563207.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000563207.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000563207.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000563207.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000564387.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000564387.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000564387.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000564387.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000564387.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000564387.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000565151.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000565151.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000565151.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000565151.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000565662.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000565662.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000565662.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000565662.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000565662.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000565662.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000565662.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000565662.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000565662.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000565662.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000566082.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000566082.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000566082.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000566082.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000566082.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000566082.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000566082.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000566082.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000566082.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000566082.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000566292.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000566292.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000566292.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000566292.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000566292.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000566292.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000566292.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000566292.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000568848.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000568848.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000568848.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000568848.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000568848.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000568848.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000568848.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000568848.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000569252.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000569252.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000569252.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000569252.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000569252.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000569252.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000569252.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000624720.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000624720.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000624720.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000624720.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000624720.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000624720.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
ENST00000624720.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
NM_001195302.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
NM_001195302.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
NM_001195302.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
NM_001195302.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
NM_001195302.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
NM_001195302.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
NM_001195302.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
NM_001195302.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
NM_001195302.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
NM_001195302.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
NM_001195302.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
NM_001195302.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
NM_001195302.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
NM_001195302.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
NM_001195302.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
NM_001195302.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
NM_001195302.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
NM_001204911.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
NM_001204911.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
NM_001204911.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
NM_001204911.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
NM_001204911.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
NM_001204911.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
NM_001204911.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
NM_001204911.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
NM_001204911.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
NM_001204911.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
NM_001204911.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
NM_001204911.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
NM_001204911.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
NM_001204911.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
NM_024598.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
NM_024598.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
NM_024598.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
NM_024598.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
NM_024598.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
NM_024598.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
NM_024598.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
NM_024598.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
NM_024598.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
NM_024598.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
NM_024598.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
NM_024598.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
NM_024598.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
NM_024598.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
NM_024598.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
NM_024598.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008020.hg.1 |
NM_024598.3 |
Refseq |