Assay Details
| Gene Symbol: USB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195302.1 | NP_001182231.1 | 4 - 5 | 673 | 88 | |
| NM_024598.3 | NP_078874.2 | 5 - 6 | 727 | 88 | ||
| XM_005256144.3 | XP_005256201.1 | 5 - 6 | 602 | 88 | ||
| XM_011523328.2 | XP_011521630.1 | 5 - 6 | 703 | 88 | ||
| XM_011523329.2 | XP_011521631.1 | 5 - 6 | 585 | 88 | ||
| GenBank mRNA | AK023216.1 | - | 5 - 6 | 88 | ||
| AK126223.1 | - | 1 - 2 | 2334 | 88 | ||
| AK293220.1 | - | 5 - 6 | 537 | 88 | ||
| AK298473.1 | - | 6 - 7 | 574 | 88 | ||
| AK303121.1 | - | 4 - 5 | 628 | 88 | ||
| BC004415.1 | - | 5 - 6 | 663 | 3641317 | 88 | |
| BC006291.2 | - | 5 - 6 | 653 | 4025993 | 88 | |
| BC007774.2 | - | 5 - 6 | 657 | 4302289 | 88 | |
| BC021554.1 | - | 5 - 6 | 647 | 4541229 | 88 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16000499.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000219281.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000219281.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000219281.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000219281.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000219281.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000219281.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000219281.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000219281.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000219281.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000219281.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000219281.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000219281.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000219281.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000219281.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000219281.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000219281.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000219281.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000423271.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000423271.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000423271.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000423271.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000423271.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000423271.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000423271.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000423271.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000423271.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000423271.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000423271.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000539737.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000539737.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000539737.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000539737.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000539737.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000539737.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000539737.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000539737.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000539737.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000539737.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000539737.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000539737.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000539737.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000539737.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000539737.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000561568.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000561568.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000561568.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000561568.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000561568.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000561568.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000561568.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000561568.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000561568.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000561743.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000561743.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000561743.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000561743.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000561743.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000561743.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000561743.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000561743.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000561743.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000561743.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000561743.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000562534.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000562534.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000562534.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000562534.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000562534.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000562534.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000562534.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000563149.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000563149.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000563149.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000563149.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000563149.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000563149.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000563149.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000563149.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000563207.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000563207.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000563207.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000563207.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000563207.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000563207.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000563207.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000563207.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000564387.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000564387.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000564387.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000564387.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000564387.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000564387.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000565151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000565151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000565151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000565151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000565662.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000565662.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000565662.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000565662.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000565662.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000565662.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000565662.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000565662.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000565662.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000565662.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000566082.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000566082.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000566082.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000566082.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000566082.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000566082.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000566082.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000566082.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000566082.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000566082.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000566292.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000566292.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000566292.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000566292.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000566292.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000566292.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000566292.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000566292.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000568848.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000568848.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000568848.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000568848.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000568848.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000568848.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000568848.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000568848.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000569252.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000569252.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000569252.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000569252.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000569252.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000569252.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000569252.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000624720.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000624720.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000624720.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000624720.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000624720.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000624720.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | ENST00000624720.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | NM_001195302.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | NM_001195302.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | NM_001195302.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | NM_001195302.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | NM_001195302.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | NM_001195302.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | NM_001195302.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | NM_001195302.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | NM_001195302.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | NM_001195302.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | NM_001195302.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | NM_001195302.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | NM_001195302.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | NM_001195302.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | NM_001195302.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | NM_001195302.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | NM_001195302.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | NM_001204911.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | NM_001204911.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | NM_001204911.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | NM_001204911.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | NM_001204911.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | NM_001204911.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | NM_001204911.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | NM_001204911.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | NM_001204911.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | NM_001204911.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | NM_001204911.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | NM_001204911.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | NM_001204911.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | NM_001204911.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | NM_024598.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | NM_024598.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | NM_024598.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | NM_024598.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | NM_024598.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | NM_024598.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | NM_024598.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | NM_024598.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | NM_024598.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | NM_024598.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | NM_024598.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | NM_024598.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | NM_024598.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | NM_024598.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | NM_024598.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | NM_024598.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008020.hg.1 | NM_024598.3 | Refseq |

USB1
Genomic Map