Assay Details
| Gene Symbol: MYL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079420.2 | NP_524144.1 | 5 - 6 | 685 | 89 | |
| NM_079422.2 | NP_524146.1 | 5 - 6 | 495 | 89 | ||
| GenBank mRNA | BC005318.1 | - | 5 - 6 | 495 | 3933998 | 89 |
| M20643.1 | - | 5 - 6 | 472 | 89 | ||
| X05451.1 | - | 5 - 6 | 477 | 89 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02002732.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR02043866.hg.1 | TC02002732.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02043866.hg.1 | ENST00000341685.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02043866.hg.1 | ENST00000352451.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02043866.hg.1 | ENST00000496436.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02043866.hg.1 | NM_079420.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02043866.hg.1 | NM_079422.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02043865.hg.1 | TC02002732.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02043865.hg.1 | ENST00000341685.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02043865.hg.1 | ENST00000352451.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02043865.hg.1 | ENST00000496436.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02043865.hg.1 | NM_079420.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02043865.hg.1 | NM_079422.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015607.hg.1 | ENST00000341685.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015607.hg.1 | ENST00000341685.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015607.hg.1 | ENST00000341685.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015607.hg.1 | ENST00000341685.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015607.hg.1 | ENST00000341685.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015607.hg.1 | ENST00000341685.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015607.hg.1 | ENST00000341685.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015607.hg.1 | ENST00000352451.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015607.hg.1 | ENST00000352451.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015607.hg.1 | ENST00000352451.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015607.hg.1 | ENST00000352451.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015607.hg.1 | ENST00000352451.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015607.hg.1 | ENST00000352451.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015607.hg.1 | ENST00000352451.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015607.hg.1 | ENST00000352451.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015607.hg.1 | ENST00000484290.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015607.hg.1 | ENST00000484290.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015607.hg.1 | ENST00000484290.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015607.hg.1 | ENST00000484290.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015607.hg.1 | ENST00000484290.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015607.hg.1 | ENST00000484290.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015607.hg.1 | ENST00000484290.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015607.hg.1 | ENST00000496436.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015607.hg.1 | ENST00000496436.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015607.hg.1 | ENST00000496436.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015607.hg.1 | ENST00000496436.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015607.hg.1 | ENST00000496436.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015607.hg.1 | ENST00000496436.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015607.hg.1 | ENST00000496436.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015607.hg.1 | NM_079420.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015607.hg.1 | NM_079420.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015607.hg.1 | NM_079420.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015607.hg.1 | NM_079420.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015607.hg.1 | NM_079420.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015607.hg.1 | NM_079420.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015607.hg.1 | NM_079420.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015607.hg.1 | NM_079420.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015607.hg.1 | NM_079422.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015607.hg.1 | NM_079422.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015607.hg.1 | NM_079422.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015607.hg.1 | NM_079422.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015607.hg.1 | NM_079422.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015607.hg.1 | NM_079422.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015607.hg.1 | NM_079422.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200214468.hg.1 | ENST00000341685.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200214468.hg.1 | ENST00000352451.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200214468.hg.1 | ENST00000484290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200214468.hg.1 | ENST00000496436.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200214468.hg.1 | NM_079420.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200214468.hg.1 | NM_079422.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200214468.hg.1 | ENST00000341685.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200214468.hg.1 | ENST00000352451.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200214468.hg.1 | ENST00000484290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200214468.hg.1 | ENST00000496436.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200214468.hg.1 | NM_079420.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200214468.hg.1 | NM_079422.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200214468.hg.1 | TC0200015607.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200015607.hg.1 | ENST00000341685.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015607.hg.1 | ENST00000341685.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015607.hg.1 | ENST00000341685.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015607.hg.1 | ENST00000341685.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015607.hg.1 | ENST00000341685.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015607.hg.1 | ENST00000341685.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015607.hg.1 | ENST00000341685.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015607.hg.1 | ENST00000352451.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015607.hg.1 | ENST00000352451.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015607.hg.1 | ENST00000352451.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015607.hg.1 | ENST00000352451.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015607.hg.1 | ENST00000352451.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015607.hg.1 | ENST00000352451.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015607.hg.1 | ENST00000352451.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015607.hg.1 | ENST00000352451.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015607.hg.1 | ENST00000484290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015607.hg.1 | ENST00000484290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015607.hg.1 | ENST00000484290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015607.hg.1 | ENST00000484290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015607.hg.1 | ENST00000484290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015607.hg.1 | ENST00000484290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015607.hg.1 | ENST00000484290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015607.hg.1 | ENST00000496436.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015607.hg.1 | ENST00000496436.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015607.hg.1 | ENST00000496436.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015607.hg.1 | ENST00000496436.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015607.hg.1 | ENST00000496436.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015607.hg.1 | ENST00000496436.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015607.hg.1 | ENST00000496436.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015607.hg.1 | NM_079420.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015607.hg.1 | NM_079420.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015607.hg.1 | NM_079420.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015607.hg.1 | NM_079420.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015607.hg.1 | NM_079420.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015607.hg.1 | NM_079420.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015607.hg.1 | NM_079420.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015607.hg.1 | NM_079420.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015607.hg.1 | NM_079422.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015607.hg.1 | NM_079422.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015607.hg.1 | NM_079422.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015607.hg.1 | NM_079422.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015607.hg.1 | NM_079422.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015607.hg.1 | NM_079422.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015607.hg.1 | NM_079422.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200214467.hg.1 | ENST00000341685.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200214467.hg.1 | ENST00000352451.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200214467.hg.1 | ENST00000496436.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200214467.hg.1 | NM_079420.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200214467.hg.1 | NM_079422.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200214467.hg.1 | ENST00000341685.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200214467.hg.1 | ENST00000352451.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200214467.hg.1 | ENST00000496436.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200214467.hg.1 | NM_079420.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200214467.hg.1 | NM_079422.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200214467.hg.1 | TC0200015607.hg.1 | Clariom D TC |

MYL1
Genomic Map