Assay Details
| Gene Symbol: PTPN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145368.1 | NP_001138840.1 | 15 - 16 | 1556 | 102 | |
| NM_001145369.1 | NP_001138841.1 | 11 - 12 | 3740 | 102 | ||
| NM_001145370.1 | NP_001138842.1 | 10 - 11 | 3605 | 102 | ||
| NM_001145371.1 | NP_001138843.1 | 6 - 7 | 1174 | 102 | ||
| NM_001145372.1 | NP_001138844.1 | 5 - 6 | 1039 | 102 | ||
| NM_002829.3 | NP_002820.3 | 16 - 17 | 1691 | 102 | ||
| NR_026918.1 | 3 - 4 | 461 | 102 | |||
| XM_006717197.3 | XP_006717260.1 | 16 - 17 | 1800 | 102 | ||
| XM_006717199.3 | XP_006717262.1 | 16 - 17 | 1825 | 102 | ||
| XM_006717201.3 | XP_006717264.1 | 15 - 16 | 1632 | 102 | ||
| XM_006717202.3 | XP_006717265.1 | 13 - 14 | 1394 | 102 | ||
| XM_006717203.3 | XP_006717266.1 | 13 - 14 | 1353 | 102 | ||
| XM_006717204.3 | XP_006717267.1 | 16 - 17 | 1727 | 102 | ||
| XM_011518888.2 | XP_011517190.1 | 16 - 17 | 1761 | 102 | ||
| XM_011518889.2 | XP_011517191.1 | 16 - 17 | 2431 | 102 | ||
| XM_017014955.1 | XP_016870444.1 | 16 - 17 | 1783 | 102 | ||
| XM_017014956.1 | XP_016870445.1 | 14 - 15 | 1504 | 102 | ||
| XM_017014957.1 | XP_016870446.1 | 15 - 16 | 1592 | 102 | ||
| GenBank mRNA | AK295875.1 | - | 14 - 15 | 1476 | 102 | |
| AK296265.1 | - | 5 - 6 | 823 | 102 | ||
| AK296388.1 | - | 7 - 8 | 1100 | 102 | ||
| AK303608.1 | - | 3 - 4 | 461 | 102 | ||
| AK304434.1 | - | 15 - 16 | 1556 | 102 | ||
| AK308349.1 | - | 3 - 4 | 424 | 102 | ||
| AK316066.1 | - | 15 - 16 | 1556 | 102 | ||
| AK316118.1 | - | 5 - 6 | 965 | 102 | ||
| BC126117.1 | - | 15 - 16 | 1601 | 8991833 | 102 | |
| BC143848.1 | - | 14 - 15 | 1466 | 9052363 | 102 | |
| BX648253.1 | - | 10 - 11 | 3606 | 102 | ||
| BX648735.1 | - | 11 - 12 | 2175 | 102 | ||
| CR749204.1 | - | 4 - 5 | 618 | 102 | ||
| DQ104439.1 | - | 15 - 16 | 1579 | 102 | ||
| M64572.1 | - | 15 - 16 | 1602 | 102 | ||
| S39392.1 | - | 8 - 9 | 1002 | 102 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09001468.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK303608.1 transcripts.
Set Membership: |
|

PTPN3
Genomic Map