Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007376.hg.1 |
ENST00000519537.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007376.hg.1 |
ENST00000519537.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007376.hg.1 |
ENST00000519537.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007376.hg.1 |
ENST00000519537.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007376.hg.1 |
ENST00000519537.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007376.hg.1 |
ENST00000519537.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007376.hg.1 |
ENST00000519537.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007376.hg.1 |
ENST00000519537.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007376.hg.1 |
ENST00000523523.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007376.hg.1 |
ENST00000523523.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007376.hg.1 |
ENST00000523523.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007376.hg.1 |
NR_046245.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007376.hg.1 |
NR_046245.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007376.hg.1 |
NR_046245.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007376.hg.1 |
NR_046245.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011551.hg.1 |
ENST00000327435.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011551.hg.1 |
ENST00000327435.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011551.hg.1 |
ENST00000327435.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011551.hg.1 |
ENST00000327435.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011551.hg.1 |
ENST00000327435.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011551.hg.1 |
ENST00000327435.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011551.hg.1 |
ENST00000327435.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011551.hg.1 |
ENST00000327435.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011551.hg.1 |
ENST00000327435.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011551.hg.1 |
ENST00000327435.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011551.hg.1 |
ENST00000327435.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011551.hg.1 |
ENST00000382093.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011551.hg.1 |
ENST00000382093.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011551.hg.1 |
ENST00000382093.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011551.hg.1 |
ENST00000382093.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011551.hg.1 |
ENST00000382093.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011551.hg.1 |
ENST00000382093.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011551.hg.1 |
ENST00000415131.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011551.hg.1 |
ENST00000415131.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011551.hg.1 |
ENST00000415131.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011551.hg.1 |
NM_001306077.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011551.hg.1 |
NM_001306077.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011551.hg.1 |
NM_001306077.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011551.hg.1 |
NM_001306077.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011551.hg.1 |
NM_001306077.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011551.hg.1 |
NM_001306077.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011551.hg.1 |
NM_001306077.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02024817.hg.1 |
TC02001524.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02024817.hg.1 |
ENST00000327435.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02024817.hg.1 |
ENST00000382093.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02024817.hg.1 |
NM_018269.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200183887.hg.1 |
ENST00000382093.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200183887.hg.1 |
NM_001306077.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200183887.hg.1 |
ENST00000382093.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200183887.hg.1 |
NM_001306077.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200183887.hg.1 |
TC0200011551.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200183884.hg.1 |
ENST00000327435.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200183884.hg.1 |
ENST00000382093.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200183884.hg.1 |
NM_001306077.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200183884.hg.1 |
ENST00000327435.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200183884.hg.1 |
ENST00000382093.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200183884.hg.1 |
NM_001306077.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200183884.hg.1 |
TC0200011551.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0800007376.hg.1 |
ENST00000519537.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007376.hg.1 |
ENST00000519537.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007376.hg.1 |
ENST00000519537.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007376.hg.1 |
ENST00000519537.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007376.hg.1 |
ENST00000519537.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007376.hg.1 |
ENST00000519537.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007376.hg.1 |
ENST00000519537.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007376.hg.1 |
ENST00000519537.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007376.hg.1 |
ENST00000523523.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007376.hg.1 |
ENST00000523523.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007376.hg.1 |
ENST00000523523.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007376.hg.1 |
NR_046245.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007376.hg.1 |
NR_046245.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007376.hg.1 |
NR_046245.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007376.hg.1 |
NR_046245.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011551.hg.1 |
ENST00000327435.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011551.hg.1 |
ENST00000327435.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011551.hg.1 |
ENST00000327435.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011551.hg.1 |
ENST00000327435.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011551.hg.1 |
ENST00000327435.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011551.hg.1 |
ENST00000327435.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011551.hg.1 |
ENST00000327435.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011551.hg.1 |
ENST00000327435.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011551.hg.1 |
ENST00000327435.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011551.hg.1 |
ENST00000327435.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011551.hg.1 |
ENST00000327435.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011551.hg.1 |
ENST00000382093.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011551.hg.1 |
ENST00000382093.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011551.hg.1 |
ENST00000382093.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011551.hg.1 |
ENST00000382093.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011551.hg.1 |
ENST00000382093.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011551.hg.1 |
ENST00000382093.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011551.hg.1 |
ENST00000415131.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011551.hg.1 |
ENST00000415131.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011551.hg.1 |
ENST00000415131.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011551.hg.1 |
NM_001306077.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011551.hg.1 |
NM_001306077.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011551.hg.1 |
NM_001306077.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011551.hg.1 |
NM_001306077.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011551.hg.1 |
NM_001306077.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011551.hg.1 |
NM_001306077.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011551.hg.1 |
NM_001306077.1 |
Refseq |