Assay Details
Gene Symbol: ACOD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258406.1 | NP_001245335.1 | 3 - 4 | 469 | 88 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of antimicrobial humoral response
defense response
inflammatory response
embryo implantation
metabolic process
negative regulation of NF-kappaB transcription factor activity
negative regulation of type I interferon production
negative regulation of toll-like receptor 2 signaling pathway
negative regulation of toll-like receptor 4 signaling pathway
cellular response to interferon-beta
negative regulation of innate immune response
negative regulation of inflammatory response
cellular response to molecule of bacterial origin
cellular response to lipopolysaccharide
cellular response to interferon-gamma
cellular response to interleukin-1
cellular response to tumor necrosis factor
cellular response to progesterone stimulus
tolerance induction to lipopolysaccharide
positive regulation of reactive oxygen species metabolic process
aconitate decarboxylase activity
positive regulation of antimicrobial humoral response
defense response
inflammatory response
embryo implantation
metabolic process
negative regulation of NF-kappaB transcription factor activity
negative regulation of type I interferon production
negative regulation of toll-like receptor 2 signaling pathway
negative regulation of toll-like receptor 4 signaling pathway
cellular response to interferon-beta
negative regulation of innate immune response
negative regulation of inflammatory response
cellular response to molecule of bacterial origin
cellular response to lipopolysaccharide
cellular response to interferon-gamma
cellular response to interleukin-1
cellular response to tumor necrosis factor
cellular response to progesterone stimulus
tolerance induction to lipopolysaccharide
positive regulation of reactive oxygen species metabolic process
aconitate decarboxylase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 53414 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3319925 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | XM_001133269_at | - | - |
Affymetrix Human Gene 1.0 ST Array | 7969486 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7969483 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7969487 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7969484 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7969485 | - | - |
Affymetrix Human Gene 1.1 ST Array | 7969482 | - | - |
Affymetrix Human Transcriptome Array 2.0 | TC1300007546.hg.1 | ENST00000377462.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007546.hg.1 | ENST00000377462.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007546.hg.1 | ENST00000377462.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007546.hg.1 | ENST00000377462.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007546.hg.1 | ENST00000377462.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007546.hg.1 | ENST00000377462.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007546.hg.1 | ENST00000449753.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007546.hg.1 | ENST00000449753.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007546.hg.1 | ENST00000449753.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007546.hg.1 | ENST00000449753.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007546.hg.1 | ENST00000449753.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007546.hg.1 | NM_001258406.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007546.hg.1 | NM_001258406.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007546.hg.1 | NM_001258406.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007546.hg.1 | NM_001258406.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300007546.hg.1 | NM_001258406.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300150985.hg.1 | ENST00000377462.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300150985.hg.1 | ENST00000449753.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300150985.hg.1 | NM_001258406.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300150985.hg.1 | ENST00000377462.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300150985.hg.1 | ENST00000449753.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300150985.hg.1 | NM_001258406.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300150985.hg.1 | TC1300007546.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1300007546.hg.1 | ENST00000377462.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007546.hg.1 | ENST00000377462.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007546.hg.1 | ENST00000377462.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007546.hg.1 | ENST00000377462.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007546.hg.1 | ENST00000377462.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007546.hg.1 | ENST00000377462.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007546.hg.1 | ENST00000449753.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007546.hg.1 | ENST00000449753.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007546.hg.1 | ENST00000449753.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007546.hg.1 | ENST00000449753.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007546.hg.1 | ENST00000449753.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007546.hg.1 | NM_001258406.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007546.hg.1 | NM_001258406.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007546.hg.1 | NM_001258406.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007546.hg.1 | NM_001258406.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300007546.hg.1 | NM_001258406.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300150986.hg.1 | ENST00000377462.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300150986.hg.1 | ENST00000449753.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300150986.hg.1 | NM_001258406.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300150986.hg.1 | ENST00000377462.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300150986.hg.1 | ENST00000449753.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300150986.hg.1 | NM_001258406.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300150986.hg.1 | TC1300007546.hg.1 | Clariom D TC |
Human Genome U133 Plus 2.0 Array | 240287_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 240287_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 240287_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 240287_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 240287_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 240287_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 240287_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 240287_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 240287_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 240287_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 240287_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 240287_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 240287_at6 | - | - |
Human Genome U95 Set | 91636_at | - | - |
Human Genome U95 Set | 91636_PM_at | - | - |
Human Genome U133 Set | 240287_PM_at | - | - |
Human Genome U133 Set | 240287_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 131561 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_1838319 | - | - |
HumanHT-12 v4 Expression BeadChip | ILMN_3193271 | - | - |
HumanHT-12 V4.0 expression beadchip | ILMN_1838319 | - | - |
HumanHT-12 V4.0 expression beadchip | ILMN_3193271 | - | - |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1838319 | - | - |
Illumina HumanWG-6_V2_0_R2 | ILMN_1838319 | - | - |
