Assay Details
Gene Symbol: HTATSF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163280.1 | NP_001156752.1 | 4 - 5 | 838 | 130 | |
NM_014500.4 | NP_055315.2 | 3 - 4 | 589 | 130 | ||
XM_005262404.3 | XP_005262461.1 | 4 - 5 | 933 | 130 | ||
GenBank mRNA | AK225870.1 | - | 3 - 4 | 536 | 130 | |
AK299403.1 | - | 3 - 4 | 541 | 130 | ||
BC009896.2 | - | 3 - 4 | 589 | 3504952 | 130 | |
BT006886.1 | - | 3 - 4 | 416 | 130 | ||
U76992.1 | - | 3 - 4 | 473 | 130 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC0X000661.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR0X008736.hg.1 | TC0X000661.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR0X008736.hg.1 | ENST00000218364.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X008736.hg.1 | ENST00000415377.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X008736.hg.1 | ENST00000535601.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X008736.hg.1 | NM_001163280.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR0X008736.hg.1 | NM_014500.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000218364.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000218364.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000218364.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000218364.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000218364.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000218364.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000218364.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000218364.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000218364.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000218364.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000218364.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000218364.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000218364.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000425695.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000425695.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000425695.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000425695.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000425695.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000425695.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000448450.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000448450.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000448450.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000448450.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000448450.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000448450.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000448450.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000448450.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000535601.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000535601.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000535601.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000535601.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000535601.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000535601.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000535601.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000535601.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000535601.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000535601.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000535601.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000535601.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000535601.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000535601.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | NM_001163280.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | NM_001163280.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | NM_001163280.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | NM_001163280.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | NM_001163280.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | NM_001163280.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | NM_001163280.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | NM_001163280.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | NM_001163280.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | NM_001163280.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | NM_001163280.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | NM_001163280.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | NM_001163280.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | NM_001163280.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | NM_014500.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | NM_014500.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | NM_014500.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | NM_014500.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | NM_014500.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | NM_014500.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | NM_014500.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | NM_014500.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | NM_014500.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | NM_014500.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | NM_014500.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | NM_014500.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | NM_014500.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00158093.hg.1 | ENST00000218364.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00158093.hg.1 | ENST00000425695.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00158093.hg.1 | ENST00000448450.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00158093.hg.1 | ENST00000535601.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00158093.hg.1 | NM_001163280.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00158093.hg.1 | NM_014500.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00158093.hg.1 | ENST00000218364.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00158093.hg.1 | ENST00000425695.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00158093.hg.1 | ENST00000448450.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00158093.hg.1 | ENST00000535601.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00158093.hg.1 | NM_001163280.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00158093.hg.1 | NM_014500.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00158093.hg.1 | TC0X00008533.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000218364.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000218364.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000218364.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000218364.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000218364.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000218364.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000218364.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000218364.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000218364.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000218364.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000218364.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000218364.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000218364.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000425695.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000425695.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000425695.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000425695.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000425695.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000425695.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000448450.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000448450.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000448450.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000448450.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000448450.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000448450.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000448450.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000448450.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000535601.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000535601.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000535601.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000535601.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000535601.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000535601.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000535601.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000535601.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000535601.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000535601.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000535601.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000535601.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000535601.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000535601.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | NM_001163280.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | NM_001163280.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | NM_001163280.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | NM_001163280.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | NM_001163280.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | NM_001163280.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | NM_001163280.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | NM_001163280.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | NM_001163280.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | NM_001163280.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | NM_001163280.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | NM_001163280.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | NM_001163280.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | NM_001163280.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | NM_014500.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | NM_014500.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | NM_014500.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | NM_014500.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | NM_014500.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | NM_014500.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | NM_014500.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | NM_014500.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | NM_014500.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | NM_014500.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | NM_014500.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | NM_014500.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | NM_014500.4 | Refseq |
