Assay Details
| Gene Symbol: HTATSF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163280.1 | NP_001156752.1 | 6 - 7 | 1159 | 111 | |
| NM_014500.4 | NP_055315.2 | 5 - 6 | 910 | 111 | ||
| XM_005262404.3 | XP_005262461.1 | 6 - 7 | 1254 | 111 | ||
| GenBank mRNA | AB209303.1 | - | 6 - 7 | 1159 | 111 | |
| AK225870.1 | - | 5 - 6 | 857 | 111 | ||
| AK299403.1 | - | 5 - 6 | 862 | 111 | ||
| BC009896.2 | - | 5 - 6 | 910 | 3504952 | 111 | |
| BT006886.1 | - | 5 - 6 | 737 | 111 | ||
| U76992.1 | - | 5 - 6 | 794 | 111 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X000661.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR0X008738.hg.1 | TC0X000661.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X008738.hg.1 | ENST00000218364.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X008738.hg.1 | ENST00000415377.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X008738.hg.1 | ENST00000535601.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X008738.hg.1 | NM_001163280.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X008738.hg.1 | NM_014500.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000218364.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000218364.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000218364.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000218364.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000218364.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000218364.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000218364.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000218364.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000218364.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000218364.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000218364.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000218364.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000218364.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000425695.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000425695.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000425695.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000425695.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000425695.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000425695.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000448450.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000448450.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000448450.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000448450.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000448450.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000448450.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000448450.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000448450.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000535601.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000535601.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000535601.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000535601.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000535601.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000535601.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000535601.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000535601.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000535601.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000535601.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000535601.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000535601.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000535601.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000535601.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | NM_001163280.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | NM_001163280.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | NM_001163280.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | NM_001163280.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | NM_001163280.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | NM_001163280.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | NM_001163280.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | NM_001163280.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | NM_001163280.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | NM_001163280.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | NM_001163280.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | NM_001163280.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | NM_001163280.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | NM_001163280.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | NM_014500.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | NM_014500.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | NM_014500.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | NM_014500.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | NM_014500.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | NM_014500.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | NM_014500.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | NM_014500.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | NM_014500.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | NM_014500.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | NM_014500.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | NM_014500.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | NM_014500.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X008739.hg.1 | TC0X000661.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X008739.hg.1 | ENST00000218364.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X008739.hg.1 | ENST00000415377.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X008739.hg.1 | ENST00000535601.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X008739.hg.1 | NM_001163280.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X008739.hg.1 | NM_014500.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00158095.hg.1 | ENST00000218364.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00158095.hg.1 | ENST00000448450.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00158095.hg.1 | ENST00000535601.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00158095.hg.1 | NM_001163280.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00158095.hg.1 | NM_014500.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00158095.hg.1 | ENST00000218364.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00158095.hg.1 | ENST00000448450.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00158095.hg.1 | ENST00000535601.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00158095.hg.1 | NM_001163280.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00158095.hg.1 | NM_014500.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00158095.hg.1 | TC0X00008533.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00158096.hg.1 | ENST00000218364.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00158096.hg.1 | ENST00000535601.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00158096.hg.1 | NM_001163280.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00158096.hg.1 | NM_014500.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00158096.hg.1 | ENST00000218364.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00158096.hg.1 | ENST00000535601.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00158096.hg.1 | NM_001163280.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00158096.hg.1 | NM_014500.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00158096.hg.1 | TC0X00008533.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000218364.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000218364.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000218364.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000218364.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000218364.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000218364.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000218364.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000218364.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000218364.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000218364.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000218364.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000218364.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000218364.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000425695.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000425695.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000425695.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000425695.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000425695.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000425695.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000448450.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000448450.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000448450.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000448450.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000448450.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000448450.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000448450.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000448450.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000535601.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000535601.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000535601.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000535601.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000535601.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000535601.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000535601.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000535601.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000535601.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000535601.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000535601.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000535601.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000535601.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000535601.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | NM_001163280.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | NM_001163280.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | NM_001163280.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | NM_001163280.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | NM_001163280.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | NM_001163280.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | NM_001163280.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | NM_001163280.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | NM_001163280.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | NM_001163280.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | NM_001163280.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | NM_001163280.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | NM_001163280.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | NM_001163280.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | NM_014500.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | NM_014500.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | NM_014500.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | NM_014500.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | NM_014500.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | NM_014500.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | NM_014500.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | NM_014500.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | NM_014500.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | NM_014500.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | NM_014500.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | NM_014500.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | NM_014500.4 | Refseq |

HTATSF1
Genomic Map