Assay Details
| Gene Symbol: HTATSF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163280.1 | NP_001156752.1 | 8 - 9 | 1350 | 80 | |
| NM_014500.4 | NP_055315.2 | 7 - 8 | 1101 | 80 | ||
| XM_005262404.3 | XP_005262461.1 | 8 - 9 | 1445 | 80 | ||
| GenBank mRNA | AB209303.1 | - | 8 - 9 | 1350 | 80 | |
| AK225870.1 | - | 7 - 8 | 1048 | 80 | ||
| AK299403.1 | - | 7 - 8 | 1053 | 80 | ||
| BC009896.2 | - | 7 - 8 | 1101 | 3504952 | 80 | |
| BT006886.1 | - | 7 - 8 | 928 | 80 | ||
| U76992.1 | - | 7 - 8 | 985 | 80 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X000661.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000218364.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000218364.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000218364.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000218364.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000218364.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000218364.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000218364.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000218364.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000218364.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000218364.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000218364.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000218364.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000218364.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000425695.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000425695.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000425695.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000425695.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000425695.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000425695.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000448450.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000448450.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000448450.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000448450.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000448450.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000448450.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000448450.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000448450.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000535601.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000535601.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000535601.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000535601.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000535601.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000535601.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000535601.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000535601.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000535601.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000535601.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000535601.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000535601.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000535601.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | ENST00000535601.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | NM_001163280.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | NM_001163280.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | NM_001163280.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | NM_001163280.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | NM_001163280.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | NM_001163280.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | NM_001163280.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | NM_001163280.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | NM_001163280.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | NM_001163280.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | NM_001163280.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | NM_001163280.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | NM_001163280.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | NM_001163280.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | NM_014500.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | NM_014500.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | NM_014500.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | NM_014500.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | NM_014500.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | NM_014500.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | NM_014500.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | NM_014500.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | NM_014500.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | NM_014500.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | NM_014500.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | NM_014500.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008533.hg.1 | NM_014500.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X008741.hg.1 | TC0X000661.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X008741.hg.1 | ENST00000218364.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X008741.hg.1 | ENST00000415377.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X008741.hg.1 | ENST00000535601.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X008741.hg.1 | NM_001163280.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X008741.hg.1 | NM_014500.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000218364.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000218364.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000218364.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000218364.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000218364.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000218364.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000218364.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000218364.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000218364.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000218364.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000218364.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000218364.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000218364.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000425695.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000425695.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000425695.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000425695.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000425695.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000425695.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000448450.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000448450.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000448450.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000448450.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000448450.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000448450.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000448450.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000448450.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000535601.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000535601.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000535601.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000535601.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000535601.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000535601.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000535601.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000535601.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000535601.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000535601.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000535601.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000535601.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000535601.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | ENST00000535601.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | NM_001163280.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | NM_001163280.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | NM_001163280.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | NM_001163280.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | NM_001163280.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | NM_001163280.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | NM_001163280.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | NM_001163280.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | NM_001163280.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | NM_001163280.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | NM_001163280.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | NM_001163280.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | NM_001163280.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | NM_001163280.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | NM_014500.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | NM_014500.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | NM_014500.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | NM_014500.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | NM_014500.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | NM_014500.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | NM_014500.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | NM_014500.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | NM_014500.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | NM_014500.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | NM_014500.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | NM_014500.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008533.hg.1 | NM_014500.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00158098.hg.1 | ENST00000218364.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00158098.hg.1 | ENST00000535601.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00158098.hg.1 | NM_001163280.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00158098.hg.1 | NM_014500.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00158098.hg.1 | ENST00000218364.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00158098.hg.1 | ENST00000535601.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00158098.hg.1 | NM_001163280.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00158098.hg.1 | NM_014500.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00158098.hg.1 | TC0X00008533.hg.1 | Clariom D TC |

HTATSF1
Genomic Map