Assay Details
| Gene Symbol: GMCL1P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_003281.1 | 1 | 2700 | 189 | ||
| GenBank mRNA | BC024184.2 | - | 1 | 2687 | 189 | |
| BC024185.1 | - | 1 | 2695 | 189 | ||
| BC033886.2 | - | 1 | 2700 | 189 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05002121.hg.1
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04489025 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24672265 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24672264 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04489029 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04489041 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04489027 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24672256 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15770591 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04489022 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04489028 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04489036 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04489042 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04489039 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04489048 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04489034 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04489044 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04489031 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04489021 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24672236 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04489038 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04489032 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24672232 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24672244 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04489033 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04489024 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04489026 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15770593 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 108209 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 8x60K | 6912 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 232681 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P425324 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_23_P425324 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 44730 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 11718 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_14_P132893 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P425324 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_23_P425324 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04489036 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15770593 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04489055 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04489034 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04489056 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04489027 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24672244 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04489035 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04489029 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24672256 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04489026 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04489042 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24672232 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04489022 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24672265 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24672267 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04489041 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04489051 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04489039 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15770591 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04489030 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04489031 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04489044 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24672236 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04489024 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04489053 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04489032 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24672264 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04489036 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04489035 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24672264 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04489024 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04489034 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24672265 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04489039 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04489030 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04489027 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24672236 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15770591 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24672244 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15770593 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04489022 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04489031 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04489042 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04489032 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04489044 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04489041 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24672232 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04489051 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24672256 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04489056 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04489026 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04489029 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04489053 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04489055 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24672267 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_23_P425324 | - | - |
| Agilent Human Genome CGH Microarray 4x44K | A_14_P132893 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P425324 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04489049 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04489023 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04489041 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04489008 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15770591 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04489061 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04489064 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04489012 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04489010 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15770593 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04489026 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04489044 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15770572 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04489031 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04489028 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04489047 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04489062 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04489024 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15770587 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04489059 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04489053 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15770590 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04489036 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15770594 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_14_P132893 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | NR_003281_at | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8116176 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8116175 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8116174 | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR05029434.hg.1 | TC05002121.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05029434.hg.1 | NR_003281.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500190963.hg.1 | NR_003281.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500190963.hg.1 | NR_003281.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500190963.hg.1 | TC0500013011.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500190964.hg.1 | ENST00000463439.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500190964.hg.1 | NR_003281.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500190964.hg.1 | ENST00000463439.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500190964.hg.1 | NR_003281.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500190964.hg.1 | TC0500013011.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500190965.hg.1 | NR_003281.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500190965.hg.1 | NR_003281.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500190965.hg.1 | TC0500013011.hg.1 | Clariom D TC |
| Human Genome U133 Plus 2.0 Array | 234488_s_at | - | - |
| Human Genome U133 Plus 2.0 Array | 234488_PM_s_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 234488_s_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 234488_s_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 234488_s_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 234488_s_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 234488_s_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 234488_s_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 234488_s_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 234488_s_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 234488_s_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 234488_s_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 234488_s_at11 | - | - |
| Human Genome U95 Set | 86673_PM_at | - | - |
| Human Genome U95 Set | 86673_at | - | - |
| Human Genome U133 Set | 234488_PM_s_at | - | - |
| Human Genome U133 Set | 234488_s_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A027761 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A027762 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A027762 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A027761 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1778105 | BC024185.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1778105 | BC033886.2 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1778105 | BC024184.2 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1778105 | NR_003281.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1731375 | - | - |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1778105 | BC024185.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1778105 | BC033886.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1778105 | BC024184.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1778105 | NR_003281.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1731375 | - | - |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1731375 | - | - |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1778105 | BC024185.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1778105 | BC033886.2 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1778105 | BC024184.2 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1778105 | NR_003281.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1778105 | BC024185.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1778105 | BC033886.2 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1778105 | BC024184.2 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1778105 | NR_003281.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1778105 | BC024185.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1778105 | BC033886.2 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1778105 | BC024184.2 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1778105 | NR_003281.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1731375 | - | - |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1778105 | BC024185.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1778105 | BC033886.2 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1778105 | BC024184.2 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1778105 | NR_003281.1 | human_transcriptome |

GMCL1P1
Genomic Map