Assay Details
Gene Symbol: SLC12A7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_006598.2 | NP_006589.2 | 14 - 15 | 1915 | 62 | |
XM_005248231.3 | XP_005248288.1 | 14 - 15 | 1920 | 62 | ||
XM_011513939.2 | XP_011512241.1 | 14 - 15 | 2009 | 62 | ||
XM_011513940.2 | XP_011512242.1 | 14 - 15 | 1844 | 62 | ||
XM_011513941.2 | XP_011512243.2 | 14 - 15 | 1939 | 62 | ||
XM_017008958.1 | XP_016864447.1 | 14 - 15 | 2219 | 62 | ||
XM_017008959.1 | XP_016864448.1 | 14 - 15 | 1871 | 62 | ||
GenBank mRNA | AF105365.1 | - | 14 - 15 | 1853 | 62 | |
AK024493.1 | - | 7 - 8 | 940 | 62 | ||
AK024497.1 | - | 7 - 8 | 765 | 62 | ||
BC098390.1 | - | 14 - 15 | 1893 | 5265743 | 62 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC05001102.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR05016011.hg.1 | TC05001102.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR05016011.hg.1 | ENST00000264930.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05016011.hg.1 | NM_006598.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR05016013.hg.1 | TC05001102.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR05016013.hg.1 | ENST00000264930.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05016013.hg.1 | NM_006598.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000504576.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000504576.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000504576.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000504576.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000504576.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000510943.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000510943.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000510943.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000510943.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000513223.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000513223.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000513223.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000513223.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000513223.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000513223.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000513223.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000513223.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000513223.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000514994.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000514994.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000514994.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000514994.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000514994.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | NM_006598.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | NM_006598.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | NM_006598.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | NM_006598.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | NM_006598.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | NM_006598.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | NM_006598.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | NM_006598.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | NM_006598.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | NM_006598.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | NM_006598.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | NM_006598.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | NM_006598.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | NM_006598.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | NM_006598.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | NM_006598.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | NM_006598.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | NM_006598.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | NM_006598.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | NM_006598.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | NM_006598.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | NM_006598.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | NM_006598.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | NM_006598.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | NM_006598.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | NM_006598.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | NM_006598.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | NM_006598.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | NM_006598.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | NM_006598.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500169429.hg.1 | ENST00000264930.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500169429.hg.1 | ENST00000504576.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500169429.hg.1 | NM_006598.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500169429.hg.1 | ENST00000264930.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500169429.hg.1 | ENST00000504576.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500169429.hg.1 | NM_006598.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500169429.hg.1 | TC0500009837.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000504576.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000504576.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000504576.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000504576.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000504576.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000510943.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000510943.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000510943.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000510943.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000513223.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000513223.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000513223.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000513223.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000513223.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000513223.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000513223.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000513223.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000513223.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000514994.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000514994.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000514994.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000514994.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000514994.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | NM_006598.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | NM_006598.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | NM_006598.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | NM_006598.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | NM_006598.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | NM_006598.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | NM_006598.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | NM_006598.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | NM_006598.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | NM_006598.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | NM_006598.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | NM_006598.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | NM_006598.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | NM_006598.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | NM_006598.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | NM_006598.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | NM_006598.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | NM_006598.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | NM_006598.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | NM_006598.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | NM_006598.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | NM_006598.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | NM_006598.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | NM_006598.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | NM_006598.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | NM_006598.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | NM_006598.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | NM_006598.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | NM_006598.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | NM_006598.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500169427.hg.1 | ENST00000264930.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500169427.hg.1 | ENST00000504576.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500169427.hg.1 | NM_006598.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500169427.hg.1 | ENST00000264930.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500169427.hg.1 | ENST00000504576.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500169427.hg.1 | NM_006598.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500169427.hg.1 | TC0500009837.hg.1 | Clariom D TC |
