Assay Details
| Gene Symbol: SLC12A7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006598.2 | NP_006589.2 | 6 - 7 | 748 | 96 | |
| XM_005248231.3 | XP_005248288.1 | 6 - 7 | 753 | 96 | ||
| XM_011513939.2 | XP_011512241.1 | 6 - 7 | 842 | 96 | ||
| XM_011513940.2 | XP_011512242.1 | 6 - 7 | 677 | 96 | ||
| XM_011513941.2 | XP_011512243.2 | 6 - 7 | 772 | 96 | ||
| XM_017008958.1 | XP_016864447.1 | 6 - 7 | 1052 | 96 | ||
| XM_017008959.1 | XP_016864448.1 | 6 - 7 | 704 | 96 | ||
| GenBank mRNA | AF105365.1 | - | 6 - 7 | 686 | 96 | |
| BC098390.1 | - | 6 - 7 | 726 | 5265743 | 96 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05001102.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000504576.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000504576.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000504576.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000504576.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000504576.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000510943.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000510943.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000510943.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000510943.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000513223.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000513223.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000513223.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000513223.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000513223.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000513223.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000513223.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000513223.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000513223.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000514994.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000514994.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000514994.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000514994.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | ENST00000514994.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05016025.hg.1 | TC05001102.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05016025.hg.1 | ENST00000264930.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05016025.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05016026.hg.1 | TC05001102.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05016026.hg.1 | ENST00000264930.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05016026.hg.1 | ENST00000343658.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05016026.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500169440.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500169440.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500169440.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500169440.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500169440.hg.1 | TC0500009837.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500169439.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500169439.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500169439.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500169439.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500169439.hg.1 | TC0500009837.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000264930.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000504576.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000504576.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000504576.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000504576.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000504576.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000510943.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000510943.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000510943.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000510943.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000513223.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000513223.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000513223.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000513223.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000513223.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000513223.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000513223.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000513223.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000513223.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000514994.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000514994.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000514994.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000514994.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | ENST00000514994.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | NM_006598.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009837.hg.1 | NM_006598.2 | Refseq |

SLC12A7
Genomic Map