Assay Details
| Gene Symbol: SETD1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014712.2 | NP_055527.1 | 9 - 10 | 3366 | 89 | |
| XM_005255723.1 | XP_005255780.1 | 9 - 10 | 2850 | 89 | ||
| XM_006721106.3 | XP_006721169.1 | 9 - 10 | 2771 | 89 | ||
| XM_017023909.1 | XP_016879398.1 | 9 - 10 | 4003 | 89 | ||
| GenBank mRNA | AB002337.2 | - | 9 - 10 | 3366 | 89 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16000364.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR16005304.hg.1 | ENST00000262519.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16005304.hg.1 | NM_014712.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16005304.hg.1 | TC16000364.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007504.hg.1 | ENST00000262519.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007504.hg.1 | ENST00000262519.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007504.hg.1 | ENST00000262519.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007504.hg.1 | ENST00000262519.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007504.hg.1 | ENST00000262519.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007504.hg.1 | ENST00000262519.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007504.hg.1 | ENST00000262519.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007504.hg.1 | ENST00000262519.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007504.hg.1 | ENST00000262519.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007504.hg.1 | ENST00000262519.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007504.hg.1 | ENST00000262519.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007504.hg.1 | ENST00000262519.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007504.hg.1 | ENST00000262519.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007504.hg.1 | ENST00000262519.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007504.hg.1 | ENST00000262519.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007504.hg.1 | ENST00000262519.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007504.hg.1 | ENST00000262519.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007504.hg.1 | ENST00000262519.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007504.hg.1 | ENST00000262519.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007504.hg.1 | ENST00000262519.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007504.hg.1 | ENST00000262519.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007504.hg.1 | ENST00000262519.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007504.hg.1 | ENST00000262519.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007504.hg.1 | ENST00000262519.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007504.hg.1 | ENST00000452917.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007504.hg.1 | ENST00000452917.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007504.hg.1 | ENST00000452917.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007504.hg.1 | ENST00000452917.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007504.hg.1 | ENST00000452917.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007504.hg.1 | ENST00000452917.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007504.hg.1 | NM_014712.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007504.hg.1 | NM_014712.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007504.hg.1 | NM_014712.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007504.hg.1 | NM_014712.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007504.hg.1 | NM_014712.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007504.hg.1 | NM_014712.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007504.hg.1 | NM_014712.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007504.hg.1 | NM_014712.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007504.hg.1 | NM_014712.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007504.hg.1 | NM_014712.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007504.hg.1 | NM_014712.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007504.hg.1 | NM_014712.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007504.hg.1 | NM_014712.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007504.hg.1 | NM_014712.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007504.hg.1 | NM_014712.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007504.hg.1 | NM_014712.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007504.hg.1 | NM_014712.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007504.hg.1 | NM_014712.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007504.hg.1 | NM_014712.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007504.hg.1 | NM_014712.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007504.hg.1 | NM_014712.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007504.hg.1 | NM_014712.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007504.hg.1 | NM_014712.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007504.hg.1 | ENST00000262519.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007504.hg.1 | ENST00000262519.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007504.hg.1 | ENST00000262519.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007504.hg.1 | ENST00000262519.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007504.hg.1 | ENST00000262519.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007504.hg.1 | ENST00000262519.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007504.hg.1 | ENST00000262519.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007504.hg.1 | ENST00000262519.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007504.hg.1 | ENST00000262519.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007504.hg.1 | ENST00000262519.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007504.hg.1 | ENST00000262519.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007504.hg.1 | ENST00000262519.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007504.hg.1 | ENST00000262519.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007504.hg.1 | ENST00000262519.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007504.hg.1 | ENST00000262519.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007504.hg.1 | ENST00000262519.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007504.hg.1 | ENST00000262519.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007504.hg.1 | ENST00000262519.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007504.hg.1 | ENST00000262519.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007504.hg.1 | ENST00000262519.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007504.hg.1 | ENST00000262519.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007504.hg.1 | ENST00000262519.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007504.hg.1 | ENST00000262519.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007504.hg.1 | ENST00000262519.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007504.hg.1 | ENST00000452917.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007504.hg.1 | ENST00000452917.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007504.hg.1 | ENST00000452917.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007504.hg.1 | ENST00000452917.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007504.hg.1 | ENST00000452917.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007504.hg.1 | ENST00000452917.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007504.hg.1 | NM_014712.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007504.hg.1 | NM_014712.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007504.hg.1 | NM_014712.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007504.hg.1 | NM_014712.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007504.hg.1 | NM_014712.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007504.hg.1 | NM_014712.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007504.hg.1 | NM_014712.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007504.hg.1 | NM_014712.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007504.hg.1 | NM_014712.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007504.hg.1 | NM_014712.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007504.hg.1 | NM_014712.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007504.hg.1 | NM_014712.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007504.hg.1 | NM_014712.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007504.hg.1 | NM_014712.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007504.hg.1 | NM_014712.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007504.hg.1 | NM_014712.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007504.hg.1 | NM_014712.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007504.hg.1 | NM_014712.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007504.hg.1 | NM_014712.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007504.hg.1 | NM_014712.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007504.hg.1 | NM_014712.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007504.hg.1 | NM_014712.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007504.hg.1 | NM_014712.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600154868.hg.1 | ENST00000262519.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600154868.hg.1 | NM_014712.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600154868.hg.1 | ENST00000262519.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600154868.hg.1 | NM_014712.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600154868.hg.1 | TC1600007504.hg.1 | Clariom D TC |

SETD1A
Genomic Map