Assay Details
Gene Symbol: LOC648987 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_034085.1 | 1 | 2304 | 66 | ||
GenBank mRNA | AK097396.1 | - | 1 | 1818 | 66 | |
BC034002.1 | - | 1 | 1314 | 66 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC05001312.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03902418 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24074356 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03902428 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15686742 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24074355 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03902465 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24074359 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24074440 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24074352 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24074456 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24074367 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03902435 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03902380 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03902398 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03902412 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03902451 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24074451 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24074354 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24074378 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03902462 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15686739 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03902446 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24074381 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24074458 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03902408 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03902454 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24074357 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24074364 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03902438 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03902400 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24074470 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03902440 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03902403 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15686737 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03902459 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24074407 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03902455 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24074468 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03902423 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03902448 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03902415 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03902407 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24074380 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03902404 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03902449 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24074460 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03902399 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03902381 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15686753 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24074467 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03902467 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24074377 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24074372 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15686741 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03902430 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24074370 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24074406 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03902431 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24074414 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03902453 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24074436 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03902424 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24074461 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03902452 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03902463 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24074408 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24074453 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03902434 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03902376 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15686744 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24074466 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03902472 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24074366 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03902402 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24074413 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24074420 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03902450 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03902442 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03902421 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03902445 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24074422 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24074363 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03902432 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03902410 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03902456 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03902405 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03902436 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03902394 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24074417 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03902429 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_24_P829181 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3331282 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_24_P829181 | - | - |
Agilent Human Genome CGH Microarray 105A (G4412A) | A_16_P37164360 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P37164360 | - | - |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 17503 | - | - |
Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 75206 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P15173418 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P37164360 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | 8111923 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8111924 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8111922 | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 227901_PM_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | PSR05018537.hg.1 | TC05001312.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR05018537.hg.1 | NR_034085.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR05018535.hg.1 | TC05001312.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR05018535.hg.1 | NR_034085.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR05018536.hg.1 | TC05001312.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR05018536.hg.1 | NR_034085.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500174176.hg.1 | NR_034085.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500174176.hg.1 | NR_034085.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500174176.hg.1 | TC0500010627.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0500174174.hg.1 | NR_034085.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500174174.hg.1 | NR_034085.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500174174.hg.1 | TC0500010627.hg.1 | Clariom D TC |
Human Genome U133 Plus 2.0 Array | 227901_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 227901_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 227901_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 227901_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 227901_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 227901_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 227901_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 227901_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 227901_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 227901_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 227901_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 227901_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 227901_at6 | - | - |
Human Genome U95 Set | 55864_at | - | - |
Human Genome U95 Set | 55864_PM_at | - | - |
Human Genome U95 Set | 87345_at | - | - |
Human Genome U95 Set | 87345_PM_at | - | - |
Human Genome U133 Set | 227901_at | - | - |
Human Genome U133 Set | 227901_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A026750 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A026750 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_1778456 | CR598822.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1778456 | AK097396.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1778456 | BC034002.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1778456 | NR_034085.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1778456 | CR598822.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1778456 | AK097396.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1778456 | BC034002.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1778456 | NR_034085.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1778456 | CR598822.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1778456 | AK097396.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1778456 | BC034002.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1778456 | NR_034085.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1778456 | CR598822.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1778456 | AK097396.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1778456 | BC034002.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1778456 | NR_034085.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1778456 | CR598822.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1778456 | AK097396.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1778456 | BC034002.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1778456 | NR_034085.1 | human_transcriptome |
