Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02047126.hg.1 |
TC02002940.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02047126.hg.1 |
ENST00000320389.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02047126.hg.1 |
ENST00000373308.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02047126.hg.1 |
ENST00000498729.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02047126.hg.1 |
NM_001244008.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02047126.hg.1 |
NM_004321.7 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200219662.hg.1 |
ENST00000320389.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200219662.hg.1 |
ENST00000404283.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200219662.hg.1 |
ENST00000431776.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200219662.hg.1 |
ENST00000498729.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200219662.hg.1 |
NM_001244008.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200219662.hg.1 |
NM_004321.7 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200219662.hg.1 |
ENST00000320389.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200219662.hg.1 |
ENST00000404283.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200219662.hg.1 |
ENST00000431776.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200219662.hg.1 |
ENST00000498729.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200219662.hg.1 |
NM_001244008.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200219662.hg.1 |
NM_004321.7 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200219662.hg.1 |
TC0200016332.hg.1 |
Clariom D TC |