Assay Details
Gene Symbol: LOC100289045 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC016740.1 | - | 1 | 186 | 61 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | Rho guanine nucleotide exchange factor 28 | ARHGEF28 |
RGNEF, RIP2, p190RHOGEF |
-10.9kb | Sense | Chr. 5 - 73626158 - 73941993 on NCBI Build GRCh38 |
Downstream | long intergenic non-protein coding RNA 1335 | LINC01335 | 352.4kb | Anti-sense | Chr. 5 - 74306410 - 74308325 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04015938 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24196608 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04015986 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04015974 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24196541 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24196594 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04015925 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24196472 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04016019 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24196603 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04015992 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24196530 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04015978 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04015921 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04015990 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04015991 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04015954 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04015959 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15710718 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24196490 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04015951 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24196591 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15710737 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24196473 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24196533 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04016030 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24196550 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24196520 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04015927 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04016044 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04015929 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04015975 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24196615 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04016025 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24196598 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04016036 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04016029 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24196596 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15710723 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04015969 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24196583 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24196495 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24196538 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04015950 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04015981 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24196524 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24196607 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24196522 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04016023 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04016045 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24196590 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24196602 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24196525 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04015960 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24196521 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04015977 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15710735 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24196595 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04016018 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04015988 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04016013 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24196589 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04015945 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24196561 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04015940 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04015983 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15710730 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24196614 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24196592 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04015946 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04015932 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15710715 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24196573 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04016017 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24196526 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24196559 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04015987 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24196556 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24196492 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24196612 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04016002 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24196513 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24196552 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24196540 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04016032 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04015953 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04015924 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24196613 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24196482 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04016000 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04016014 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04015948 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04016011 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04015943 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04015979 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04015935 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04016016 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24196536 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24196486 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24196578 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04015934 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24196588 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04015961 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24196509 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24196558 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04015957 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04016028 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24196551 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24196549 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15710736 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04015944 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24196562 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04015989 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 578947 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_24_P93874 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 43993 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_24_P93874 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P01257231 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P93874 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P01257231 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P01257231 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix HT HG-U133+ PM Array Plate | 1564749_PM_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | TC0500007782.hg.1 | ENST00000565306.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500007782.hg.1 | ENST00000599835.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007782.hg.1 | ENST00000565306.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500007782.hg.1 | ENST00000599835.1 | Ensembl |
Human Genome U133 Plus 2.0 Array | 1564749_at | - | - |
Human Genome U133 Plus 2.0 Array | 1564749_PM_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1564749_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1564749_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1564749_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1564749_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1564749_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1564749_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1564749_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1564749_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1564749_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1564749_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1564749_at5 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 229519 | - | - |
