Assay Details

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the AB026049.1,AK298541.1 transcripts.
This assay is in the 5'UTR region of the AB026049.1,AK298541.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
