Assay Details
| Gene Symbol: SS18L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301778.1 | NP_001288707.1 | 3 - 4 | 431 | 60 | |
| NM_198935.2 | NP_945173.1 | 2 - 3 | 246 | 60 | ||
| NR_125981.1 | 2 - 3 | 246 | 60 | |||
| XM_005260389.3 | XP_005260446.1 | 2 - 3 | 201 | 60 | ||
| XM_005260391.1 | XP_005260448.1 | 3 - 4 | 385 | 60 | ||
| XM_011528764.2 | XP_011527066.1 | 3 - 4 | 1377 | 60 | ||
| XM_017027784.1 | XP_016883273.1 | 2 - 3 | 567 | 60 | ||
| GenBank mRNA | AB014593.1 | - | 2 - 3 | 171 | 60 | |
| AK125656.1 | - | 3 - 4 | 406 | 60 | ||
| AK291485.1 | - | 2 - 3 | 221 | 60 | ||
| AK299881.1 | - | 2 - 3 | 221 | 60 | ||
| BC034494.2 | - | 2 - 3 | 200 | 4823298 | 60 | |
| BX640848.1 | - | 3 - 4 | 383 | 60 | ||
| DC375156.1 | - | 2 - 3 | 246 | 60 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC20000483.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK125656.1,NM_001301778.1 transcripts.
Set Membership: |
|

SS18L1
Genomic Map