Assay Details
| Gene Symbol: UCHL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006002.4 | NP_005993.1 | 1 - 2 | 114 | 75 | |
| XM_011535212.1 | XP_011533514.1 | 1 - 2 | 114 | 75 | ||
| XM_017020725.1 | XP_016876214.1 | 1 - 2 | 114 | 75 | ||
| XM_017020726.1 | XP_016876215.1 | 1 - 2 | 114 | 75 | ||
| GenBank mRNA | AK313665.1 | - | 1 - 2 | 104 | 75 | |
| BC018125.2 | - | 1 - 2 | 73 | 3887201 | 75 | |
| BT019359.1 | - | 1 - 2 | 43 | 75 | ||
| CR456855.1 | - | 1 - 2 | 43 | 75 | ||
| DA820150.1 | - | 1 - 2 | 114 | 75 | ||
| M30496.1 | - | 1 - 2 | 63 | 75 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC13000285.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | ENST00000377595.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | ENST00000377595.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | ENST00000377595.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | ENST00000377595.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | ENST00000377595.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | ENST00000377595.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | ENST00000377595.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | ENST00000377595.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | ENST00000377595.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | ENST00000377595.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | ENST00000377595.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | ENST00000419068.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | ENST00000419068.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | ENST00000419068.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | ENST00000419068.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | ENST00000419068.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | ENST00000419068.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | ENST00000419068.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | ENST00000419068.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | ENST00000471792.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | ENST00000471792.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | ENST00000471792.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | ENST00000471792.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | ENST00000471792.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | ENST00000471792.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | ENST00000471792.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | ENST00000606347.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | ENST00000606347.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | ENST00000606347.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | ENST00000606347.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | ENST00000606347.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | ENST00000606347.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | ENST00000607339.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | ENST00000607339.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | ENST00000607339.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | ENST00000607339.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | ENST00000618773.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | ENST00000618773.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | ENST00000618773.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | ENST00000618773.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | ENST00000618773.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | ENST00000618773.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | ENST00000618773.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | ENST00000618773.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | ENST00000618773.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | ENST00000618773.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | ENST00000618773.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | NM_001270952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | NM_001270952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | NM_001270952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | NM_001270952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | NM_001270952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | NM_001270952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | NM_001270952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | NM_001270952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | NM_001270952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | NM_001270952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | NM_001270952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | NM_006002.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | NM_006002.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | NM_006002.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | NM_006002.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | NM_006002.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | NM_006002.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | NM_006002.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | NM_006002.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | NM_006002.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | NM_006002.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | NM_006002.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | NM_006002.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | NM_006002.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13003031.hg.1 | ENST00000377589.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13003031.hg.1 | ENST00000377595.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13003031.hg.1 | ENST00000471792.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13003031.hg.1 | NM_006002.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13003031.hg.1 | TC13000285.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR13003029.hg.1 | ENST00000377589.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13003029.hg.1 | ENST00000377595.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13003029.hg.1 | NM_006002.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13003029.hg.1 | TC13000285.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1300166706.hg.1 | ENST00000377595.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300166706.hg.1 | NM_006002.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300166706.hg.1 | ENST00000377595.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300166706.hg.1 | NM_006002.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300166706.hg.1 | TC1300009979.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | ENST00000377595.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | ENST00000377595.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | ENST00000377595.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | ENST00000377595.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | ENST00000377595.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | ENST00000377595.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | ENST00000377595.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | ENST00000377595.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | ENST00000377595.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | ENST00000377595.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | ENST00000377595.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | ENST00000419068.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | ENST00000419068.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | ENST00000419068.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | ENST00000419068.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | ENST00000419068.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | ENST00000419068.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | ENST00000419068.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | ENST00000419068.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | ENST00000471792.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | ENST00000471792.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | ENST00000471792.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | ENST00000471792.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | ENST00000471792.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | ENST00000471792.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | ENST00000471792.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | ENST00000606347.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | ENST00000606347.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | ENST00000606347.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | ENST00000606347.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | ENST00000606347.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | ENST00000606347.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | ENST00000607339.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | ENST00000607339.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | ENST00000607339.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | ENST00000607339.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | ENST00000618773.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | ENST00000618773.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | ENST00000618773.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | ENST00000618773.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | ENST00000618773.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | ENST00000618773.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | ENST00000618773.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | ENST00000618773.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | ENST00000618773.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | ENST00000618773.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | ENST00000618773.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | NM_001270952.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | NM_001270952.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | NM_001270952.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | NM_001270952.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | NM_001270952.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | NM_001270952.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | NM_001270952.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | NM_001270952.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | NM_001270952.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | NM_001270952.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | NM_001270952.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | NM_006002.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | NM_006002.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | NM_006002.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | NM_006002.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | NM_006002.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | NM_006002.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | NM_006002.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | NM_006002.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | NM_006002.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | NM_006002.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | NM_006002.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | NM_006002.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | NM_006002.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300166704.hg.1 | NM_006002.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300166704.hg.1 | NM_006002.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300166704.hg.1 | TC1300009979.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1300166705.hg.1 | ENST00000377595.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300166705.hg.1 | NM_006002.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300166705.hg.1 | ENST00000377595.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300166705.hg.1 | NM_006002.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300166705.hg.1 | TC1300009979.hg.1 | Clariom D TC |

UCHL3
Genomic Map