Assay Details
Gene Symbol: CRIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270837.1 | NP_001257766.1 | 3 - 4 | 732 | 72 | |
NM_001312.3 | NP_001303.1 | 3 - 4 | 288 | 72 | ||
NR_073082.1 | 3 - 4 | 264 | 72 | |||
NR_073083.1 | 2 - 3 | 193 | 72 | |||
NR_073084.1 | 3 - 4 | 1041 | 72 | |||
NR_073085.1 | 3 - 4 | 513 | 72 | |||
GenBank mRNA | AK057837.1 | - | 2 - 3 | 147 | 72 | |
AK091845.1 | - | 3 - 4 | 1041 | 72 | ||
AK223251.1 | - | 3 - 4 | 200 | 72 | ||
AK300092.1 | - | 3 - 4 | 648 | 72 | ||
AK311402.1 | - | 3 - 4 | 513 | 72 | ||
AK315757.1 | - | 3 - 4 | 240 | 72 | ||
BC000434.2 | - | 3 - 4 | 219 | 2819982 | 72 | |
BC001931.1 | - | 3 - 4 | 219 | 2819982 | 72 | |
BC034151.1 | - | 3 - 4 | 269 | 4690443 | 72 | |
BC128101.1 | - | 3 - 4 | 243 | 40109449 | 72 | |
BE271979.1 | - | 3 - 4 | 187 | 72 | ||
BT019911.1 | - | 3 - 4 | 200 | 72 | ||
CN367966.1 | - | 2 - 3 | 151 | 72 | ||
D42123.1 | - | 3 - 4 | 226 | 72 | ||
U36190.1 | - | 3 - 4 | 236 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
