Assay Details
| Gene Symbol: CD164 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142402.1 | NP_001135874.1 | 3 - 4 | 516 | 157 | |
| GenBank mRNA | AF106518.1 | - | 3 - 4 | 335 | 157 | |
| AF299342.1 | - | 3 - 4 | 413 | 157 | ||
| AK301692.1 | - | 3 - 4 | 426 | 157 | ||
| AK303525.1 | - | 3 - 4 | 368 | 157 | ||
More Information
Set Membership: |
|

CD164
Genomic Map