Assay Details
Gene Symbol: PTGER3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198714.1 | NP_942007.1 | 4 - 5 | 1424 | 82 | |
GenBank mRNA | AK290634.1 | - | 4 - 5 | 1290 | 82 | |
D38298.1 | - | 4 - 5 | 1424 | 82 | ||
S69326.1 | - | 4 - 5 | 1389 | 82 | ||
X83857.1 | - | 4 - 5 | 1400 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
