Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Clariom D Assay |
PSR0600159413.hg.1 |
ENST00000287097.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600159413.hg.1 |
ENST00000422508.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600159413.hg.1 |
ENST00000437994.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600159413.hg.1 |
NM_001159587.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600159413.hg.1 |
NM_001159588.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600159413.hg.1 |
NM_133493.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600159413.hg.1 |
ENST00000287097.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600159413.hg.1 |
ENST00000422508.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600159413.hg.1 |
ENST00000437994.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600159413.hg.1 |
NM_001159587.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600159413.hg.1 |
NM_001159588.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600159413.hg.1 |
NM_133493.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600159413.hg.1 |
TC0600008539.hg.1 |
Clariom D TC |