Assay Details
| Gene Symbol: CD109 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159587.2 | NP_001153059.1 | 19 - 20 | 2357 | 110 | |
| NM_001159588.2 | NP_001153060.1 | 18 - 19 | 2126 | 110 | ||
| NM_133493.4 | NP_598000.2 | 19 - 20 | 2357 | 110 | ||
| XM_005248659.3 | XP_005248716.1 | 20 - 21 | 2504 | 110 | ||
| XM_011535473.1 | XP_011533775.1 | 19 - 20 | 2202 | 110 | ||
| XM_017010283.1 | XP_016865772.1 | 17 - 18 | 1969 | 110 | ||
| GenBank mRNA | AF410459.1 | - | 19 - 20 | 2338 | 110 | |
| AK123960.1 | - | 5 - 6 | 435 | 110 | ||
| AK313636.1 | - | 19 - 20 | 2337 | 110 | ||
| AL834478.1 | - | 5 - 6 | 453 | 110 | ||
| AY149920.1 | - | 19 - 20 | 2226 | 110 | ||
| AY788891.1 | - | 19 - 20 | 2251 | 110 | ||
| BX641095.1 | - | 18 - 19 | 2013 | 110 | ||
| EF553520.1 | - | 19 - 20 | 2256 | 110 | ||
More Information
Set Membership: |
|

CD109
Genomic Map