Assay Details
| Gene Symbol: MSH3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002439.4 | NP_002430.3 | 13 - 14 | 1978 | 149 | |
| GenBank mRNA | AK289856.1 | - | 14 - 15 | 1947 | 149 | |
| BC004177.2 | - | 14 - 15 | 1919 | 149 | ||
| BC011817.2 | - | 14 - 15 | 1919 | 149 | ||
| BC017273.2 | - | 13 - 14 | 1901 | 149 | ||
| BC098436.1 | - | 14 - 15 | 1919 | 149 | ||
| BC130434.1 | - | 13 - 14 | 1978 | 40146465 | 149 | |
| BC130436.1 | - | 13 - 14 | 1978 | 40146467 | 149 | |
| U61981.1 | - | 14 - 15 | 1887 | 149 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05000400.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
meiotic mismatch repair
DNA repair
mismatch repair
reciprocal meiotic recombination
maintenance of DNA repeat elements
negative regulation of DNA recombination
positive regulation of helicase activity
damaged DNA binding
single-stranded DNA binding
protein binding
ATP binding
enzyme binding
mismatched DNA binding
guanine/thymine mispair binding
dinucleotide insertion or deletion binding
single guanine insertion binding
dinucleotide repeat insertion binding
oxidized purine DNA binding
protein homodimerization activity
meiotic mismatch repair
DNA repair
mismatch repair
reciprocal meiotic recombination
maintenance of DNA repeat elements
negative regulation of DNA recombination
positive regulation of helicase activity
damaged DNA binding
single-stranded DNA binding
protein binding
ATP binding
enzyme binding
mismatched DNA binding
guanine/thymine mispair binding
dinucleotide insertion or deletion binding
single guanine insertion binding
dinucleotide repeat insertion binding
oxidized purine DNA binding
protein homodimerization activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0500007919.hg.1 | ENST00000265081.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007919.hg.1 | ENST00000265081.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007919.hg.1 | ENST00000265081.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007919.hg.1 | ENST00000265081.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007919.hg.1 | ENST00000265081.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007919.hg.1 | ENST00000265081.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007919.hg.1 | ENST00000265081.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007919.hg.1 | ENST00000265081.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007919.hg.1 | ENST00000265081.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007919.hg.1 | ENST00000265081.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007919.hg.1 | ENST00000265081.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007919.hg.1 | ENST00000265081.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007919.hg.1 | ENST00000265081.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007919.hg.1 | ENST00000265081.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007919.hg.1 | ENST00000265081.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007919.hg.1 | ENST00000265081.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007919.hg.1 | ENST00000265081.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007919.hg.1 | ENST00000265081.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007919.hg.1 | ENST00000265081.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007919.hg.1 | ENST00000265081.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007919.hg.1 | ENST00000265081.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007919.hg.1 | ENST00000265081.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007919.hg.1 | ENST00000265081.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007919.hg.1 | ENST00000265081.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007919.hg.1 | ENST00000265081.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007919.hg.1 | ENST00000265081.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007919.hg.1 | ENST00000265081.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007919.hg.1 | ENST00000265081.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007919.hg.1 | ENST00000512258.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007919.hg.1 | ENST00000512258.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007919.hg.1 | ENST00000512258.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007919.hg.1 | ENST00000512258.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007919.hg.1 | ENST00000512258.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007919.hg.1 | ENST00000512258.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007919.hg.1 | ENST00000512531.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007919.hg.1 | ENST00000512531.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007919.hg.1 | NM_002439.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007919.hg.1 | NM_002439.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007919.hg.1 | NM_002439.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007919.hg.1 | NM_002439.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007919.hg.1 | NM_002439.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007919.hg.1 | NM_002439.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007919.hg.1 | NM_002439.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007919.hg.1 | NM_002439.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007919.hg.1 | NM_002439.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007919.hg.1 | NM_002439.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007919.hg.1 | NM_002439.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007919.hg.1 | NM_002439.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007919.hg.1 | NM_002439.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007919.hg.1 | NM_002439.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007919.hg.1 | NM_002439.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007919.hg.1 | NM_002439.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007919.hg.1 | NM_002439.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007919.hg.1 | NM_002439.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007919.hg.1 | NM_002439.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007919.hg.1 | NM_002439.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007919.hg.1 | NM_002439.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007919.hg.1 | NM_002439.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007919.hg.1 | NM_002439.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007919.hg.1 | NM_002439.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007919.hg.1 | NM_002439.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007919.hg.1 | NM_002439.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007919.hg.1 | NM_002439.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007919.hg.1 | NM_002439.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007919.hg.1 | NM_002439.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05005704.hg.1 | TC05000400.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05005704.hg.1 | ENST00000265081.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05005704.hg.1 | ENST00000512258.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05005704.hg.1 | ENST00000535995.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05005704.hg.1 | NM_002439.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500154250.hg.1 | ENST00000265081.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500154250.hg.1 | NM_002439.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500154250.hg.1 | ENST00000265081.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500154250.hg.1 | NM_002439.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500154250.hg.1 | TC0500007919.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0500007919.hg.1 | ENST00000265081.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007919.hg.1 | ENST00000265081.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007919.hg.1 | ENST00000265081.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007919.hg.1 | ENST00000265081.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007919.hg.1 | ENST00000265081.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007919.hg.1 | ENST00000265081.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007919.hg.1 | ENST00000265081.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007919.hg.1 | ENST00000265081.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007919.hg.1 | ENST00000265081.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007919.hg.1 | ENST00000265081.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007919.hg.1 | ENST00000265081.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007919.hg.1 | ENST00000265081.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007919.hg.1 | ENST00000265081.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007919.hg.1 | ENST00000265081.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007919.hg.1 | ENST00000265081.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007919.hg.1 | ENST00000265081.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007919.hg.1 | ENST00000265081.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007919.hg.1 | ENST00000265081.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007919.hg.1 | ENST00000265081.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007919.hg.1 | ENST00000265081.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007919.hg.1 | ENST00000265081.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007919.hg.1 | ENST00000265081.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007919.hg.1 | ENST00000265081.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007919.hg.1 | ENST00000265081.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007919.hg.1 | ENST00000265081.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007919.hg.1 | ENST00000265081.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007919.hg.1 | ENST00000265081.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007919.hg.1 | ENST00000265081.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007919.hg.1 | ENST00000512258.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007919.hg.1 | ENST00000512258.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007919.hg.1 | ENST00000512258.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007919.hg.1 | ENST00000512258.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007919.hg.1 | ENST00000512258.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007919.hg.1 | ENST00000512258.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007919.hg.1 | ENST00000512531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007919.hg.1 | ENST00000512531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007919.hg.1 | NM_002439.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007919.hg.1 | NM_002439.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007919.hg.1 | NM_002439.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007919.hg.1 | NM_002439.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007919.hg.1 | NM_002439.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007919.hg.1 | NM_002439.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007919.hg.1 | NM_002439.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007919.hg.1 | NM_002439.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007919.hg.1 | NM_002439.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007919.hg.1 | NM_002439.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007919.hg.1 | NM_002439.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007919.hg.1 | NM_002439.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007919.hg.1 | NM_002439.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007919.hg.1 | NM_002439.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007919.hg.1 | NM_002439.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007919.hg.1 | NM_002439.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007919.hg.1 | NM_002439.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007919.hg.1 | NM_002439.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007919.hg.1 | NM_002439.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007919.hg.1 | NM_002439.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007919.hg.1 | NM_002439.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007919.hg.1 | NM_002439.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007919.hg.1 | NM_002439.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007919.hg.1 | NM_002439.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007919.hg.1 | NM_002439.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007919.hg.1 | NM_002439.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007919.hg.1 | NM_002439.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007919.hg.1 | NM_002439.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007919.hg.1 | NM_002439.4 | Refseq |

MSH3
Genomic Map