Assay Details
| Gene Symbol: CYP24A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000782.4 | NP_000773.2 | 3 - 4 | 938 | 89 | |
| NM_001128915.1 | NP_001122387.1 | 3 - 4 | 938 | 89 | ||
| XM_005260304.4 | XP_005260361.1 | 3 - 4 | 946 | 89 | ||
| XM_017027691.1 | XP_016883180.1 | 3 - 4 | 946 | 89 | ||
| XM_017027692.1 | XP_016883181.1 | 3 - 4 | 946 | 89 | ||
| XM_017027693.1 | XP_016883182.1 | 3 - 4 | 946 | 89 | ||
| GenBank mRNA | AY858838.1 | - | 1 - 2 | 374 | 89 | |
| BC109083.1 | - | 3 - 4 | 625 | 40034385 | 89 | |
| BC109084.1 | - | 3 - 4 | 625 | 40034387 | 89 | |
| L13286.1 | - | 3 - 4 | 930 | 89 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC20000962.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
osteoblast differentiation
vitamin metabolic process
response to vitamin D
vitamin D metabolic process
vitamin D catabolic process
oxidation-reduction process
vitamin D receptor signaling pathway
iron ion binding
25-hydroxycholecalciferol-24-hydroxylase activity
oxidoreductase activity
heme binding
1-alpha,25-dihydroxyvitamin D3 24-hydroxylase activity
osteoblast differentiation
vitamin metabolic process
response to vitamin D
vitamin D metabolic process
vitamin D catabolic process
oxidation-reduction process
vitamin D receptor signaling pathway
iron ion binding
25-hydroxycholecalciferol-24-hydroxylase activity
oxidoreductase activity
heme binding
1-alpha,25-dihydroxyvitamin D3 24-hydroxylase activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR20013223.hg.1 | ENST00000216862.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20013223.hg.1 | ENST00000395954.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20013223.hg.1 | ENST00000395955.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20013223.hg.1 | NM_000782.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20013223.hg.1 | NM_001128915.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20013223.hg.1 | TC20000962.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000216862.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000216862.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000216862.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000216862.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000216862.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000216862.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000216862.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000216862.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000216862.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000216862.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000216862.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000216862.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000216862.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000216862.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000216862.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000216862.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000216862.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000216862.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000395954.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000395954.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000395954.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000395954.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000395954.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000395954.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000395954.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000395954.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000395954.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000395954.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000395954.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000395955.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000395955.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000395955.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000395955.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000395955.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000395955.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000395955.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000395955.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000395955.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000395955.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000395955.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000395955.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000395955.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000395955.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000395955.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000460643.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000460643.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000460643.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000460643.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000472970.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000472970.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000472970.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000487593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000487593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000487593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | NM_000782.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | NM_000782.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | NM_000782.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | NM_000782.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | NM_000782.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | NM_000782.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | NM_000782.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | NM_000782.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | NM_000782.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | NM_000782.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | NM_000782.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | NM_000782.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | NM_000782.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | NM_000782.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | NM_000782.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | NM_000782.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | NM_000782.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | NM_000782.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | NM_001128915.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | NM_001128915.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | NM_001128915.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | NM_001128915.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | NM_001128915.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | NM_001128915.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | NM_001128915.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | NM_001128915.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | NM_001128915.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | NM_001128915.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | NM_001128915.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | NM_001128915.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | NM_001128915.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | NM_001128915.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | NM_001128915.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | NM_001128915.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | NM_001128915.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000166541.hg.1 | ENST00000216862.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000166541.hg.1 | ENST00000395954.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000166541.hg.1 | ENST00000395955.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000166541.hg.1 | NM_000782.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000166541.hg.1 | NM_001128915.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000166541.hg.1 | TC2000009522.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000166541.hg.1 | ENST00000216862.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000166541.hg.1 | ENST00000395954.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000166541.hg.1 | ENST00000395955.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000166541.hg.1 | NM_000782.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000166541.hg.1 | NM_001128915.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000216862.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000216862.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000216862.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000216862.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000216862.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000216862.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000216862.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000216862.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000216862.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000216862.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000216862.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000216862.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000216862.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000216862.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000216862.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000216862.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000216862.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000216862.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000395954.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000395954.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000395954.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000395954.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000395954.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000395954.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000395954.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000395954.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000395954.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000395954.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000395954.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000395955.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000395955.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000395955.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000395955.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000395955.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000395955.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000395955.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000395955.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000395955.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000395955.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000395955.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000395955.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000395955.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000395955.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000395955.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000460643.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000460643.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000460643.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000460643.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000472970.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000472970.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000472970.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000487593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000487593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000487593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | NM_000782.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | NM_000782.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | NM_000782.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | NM_000782.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | NM_000782.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | NM_000782.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | NM_000782.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | NM_000782.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | NM_000782.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | NM_000782.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | NM_000782.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | NM_000782.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | NM_000782.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | NM_000782.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | NM_000782.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | NM_000782.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | NM_000782.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | NM_000782.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | NM_001128915.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | NM_001128915.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | NM_001128915.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | NM_001128915.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | NM_001128915.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | NM_001128915.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | NM_001128915.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | NM_001128915.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | NM_001128915.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | NM_001128915.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | NM_001128915.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | NM_001128915.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | NM_001128915.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | NM_001128915.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | NM_001128915.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | NM_001128915.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | NM_001128915.1 | Refseq |

CYP24A1
Genomic Map