| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15012290.hg.1 |
ENST00000267812.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15012290.hg.1 |
NM_005926.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15012290.hg.1 |
TC15001287.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15012289.hg.1 |
ENST00000267812.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15012289.hg.1 |
NM_005926.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15012289.hg.1 |
TC15001287.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15012292.hg.1 |
ENST00000267812.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15012292.hg.1 |
NM_005926.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15012292.hg.1 |
TC15001287.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009236.hg.1 |
ENST00000267812.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009236.hg.1 |
ENST00000267812.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009236.hg.1 |
ENST00000267812.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009236.hg.1 |
ENST00000267812.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009236.hg.1 |
ENST00000267812.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009236.hg.1 |
ENST00000267812.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009236.hg.1 |
ENST00000267812.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009236.hg.1 |
ENST00000267812.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009236.hg.1 |
ENST00000267812.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009236.hg.1 |
ENST00000267812.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009236.hg.1 |
ENST00000267812.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009236.hg.1 |
ENST00000267812.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009236.hg.1 |
ENST00000267812.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009236.hg.1 |
ENST00000267812.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009236.hg.1 |
ENST00000267812.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009236.hg.1 |
ENST00000267812.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009236.hg.1 |
ENST00000484386.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009236.hg.1 |
ENST00000484386.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009236.hg.1 |
ENST00000484386.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009236.hg.1 |
ENST00000484386.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009236.hg.1 |
ENST00000484386.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009236.hg.1 |
NM_005926.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009236.hg.1 |
NM_005926.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009236.hg.1 |
NM_005926.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009236.hg.1 |
NM_005926.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009236.hg.1 |
NM_005926.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009236.hg.1 |
NM_005926.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009236.hg.1 |
NM_005926.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009236.hg.1 |
NM_005926.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009236.hg.1 |
NM_005926.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009236.hg.1 |
NM_005926.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009236.hg.1 |
NM_005926.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009236.hg.1 |
NM_005926.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009236.hg.1 |
NM_005926.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009236.hg.1 |
NM_005926.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008520.hg.1 |
ENST00000442257.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500167641.hg.1 |
ENST00000267812.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500167641.hg.1 |
NM_005926.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500167641.hg.1 |
ENST00000267812.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500167641.hg.1 |
NM_005926.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500167641.hg.1 |
TC1500009236.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1300008520.hg.1 |
ENST00000442257.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500167642.hg.1 |
ENST00000267812.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500167642.hg.1 |
NM_005926.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500167642.hg.1 |
ENST00000267812.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500167642.hg.1 |
NM_005926.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500167642.hg.1 |
TC1500009236.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1500167643.hg.1 |
ENST00000267812.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500167643.hg.1 |
NM_005926.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500167643.hg.1 |
ENST00000267812.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500167643.hg.1 |
NM_005926.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500167643.hg.1 |
TC1500009236.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1500009236.hg.1 |
ENST00000267812.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009236.hg.1 |
ENST00000267812.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009236.hg.1 |
ENST00000267812.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009236.hg.1 |
ENST00000267812.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009236.hg.1 |
ENST00000267812.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009236.hg.1 |
ENST00000267812.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009236.hg.1 |
ENST00000267812.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009236.hg.1 |
ENST00000267812.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009236.hg.1 |
ENST00000267812.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009236.hg.1 |
ENST00000267812.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009236.hg.1 |
ENST00000267812.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009236.hg.1 |
ENST00000267812.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009236.hg.1 |
ENST00000267812.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009236.hg.1 |
ENST00000267812.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009236.hg.1 |
ENST00000267812.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009236.hg.1 |
ENST00000267812.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009236.hg.1 |
ENST00000484386.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009236.hg.1 |
ENST00000484386.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009236.hg.1 |
ENST00000484386.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009236.hg.1 |
ENST00000484386.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009236.hg.1 |
ENST00000484386.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009236.hg.1 |
NM_005926.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500009236.hg.1 |
NM_005926.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500009236.hg.1 |
NM_005926.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500009236.hg.1 |
NM_005926.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500009236.hg.1 |
NM_005926.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500009236.hg.1 |
NM_005926.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500009236.hg.1 |
NM_005926.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500009236.hg.1 |
NM_005926.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500009236.hg.1 |
NM_005926.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500009236.hg.1 |
NM_005926.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500009236.hg.1 |
NM_005926.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500009236.hg.1 |
NM_005926.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500009236.hg.1 |
NM_005926.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500009236.hg.1 |
NM_005926.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500167645.hg.1 |
ENST00000267812.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500167645.hg.1 |
NM_005926.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500167645.hg.1 |
ENST00000267812.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500167645.hg.1 |
NM_005926.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500167645.hg.1 |
TC1500009236.hg.1 |
Clariom D TC |