Assay Details
Gene Symbol: C20orf173 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_026933.2 | 9 - 10 | 1597 | 60 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | centrosomal protein 250 | CEP250 |
C-NAP1, CEP2, CNAP1 |
-4.4kb | Anti-sense | Chr. 20 - 35455138 - 35517531 on NCBI Build GRCh38 |
Downstream | ERGIC and golgi 3 | ERGIC3 |
C20orf47, CGI-54, Erv46, NY-BR-84, PRO0989, SDBCAG84, dJ477O4.2 |
19.8kb | Anti-sense | Chr. 20 - 35542029 - 35557634 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC20000801.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR20010776.hg.1 | ENST00000422009.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20010776.hg.1 | NR_026933.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR20010776.hg.1 | TC20000801.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000246199.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000246199.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000246199.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000246199.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000246199.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000246199.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000246199.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000246199.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000246199.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000374345.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000374345.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000374345.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000374345.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000374345.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000374345.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000374345.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000374345.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000374345.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000374345.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000374345.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000422009.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000422009.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000422009.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000422009.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000422009.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000422009.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000422009.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000422009.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000422009.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000422009.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000422009.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000422009.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000424444.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000424444.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000424444.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000432650.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000432650.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000432650.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000432650.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000432650.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000432650.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000432650.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000444723.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000444723.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000444723.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000444723.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000444723.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000444723.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000444723.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000444723.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000444723.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000444723.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000444723.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000450773.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000450773.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000450773.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000450773.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000450773.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | NM_001145350.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | NM_001145350.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | NM_001145350.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | NM_001145350.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | NM_001145350.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | NM_001145350.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | NM_001145350.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | NM_001145350.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | NM_001145350.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | NM_001145350.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | NM_001145350.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | NR_026933.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | NR_026933.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | NR_026933.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | NR_026933.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | NR_026933.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | NR_026933.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | NR_026933.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | NR_026933.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | NR_026933.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | NR_026933.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | NR_026933.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | NR_026933.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | NR_026933.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | NR_026933.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | NR_026933.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | NR_026933.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | NR_026933.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | NR_026933.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | NR_026933.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | NR_026933.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | NR_026933.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | NR_026933.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR20010777.hg.1 | ENST00000422009.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20010777.hg.1 | NR_026933.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR20010777.hg.1 | TC20000801.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR2000162576.hg.1 | ENST00000422009.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000162576.hg.1 | ENST00000450773.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000162576.hg.1 | NR_026933.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000162576.hg.1 | TC2000008975.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2000162576.hg.1 | ENST00000422009.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000162576.hg.1 | ENST00000450773.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000162576.hg.1 | NR_026933.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000162579.hg.1 | ENST00000422009.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000162579.hg.1 | ENST00000432650.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000162579.hg.1 | NR_026933.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000162579.hg.1 | TC2000008975.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2000162579.hg.1 | ENST00000422009.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000162579.hg.1 | ENST00000432650.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000162579.hg.1 | NR_026933.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000246199.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000246199.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000246199.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000246199.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000246199.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000246199.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000246199.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000246199.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000246199.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000374345.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000374345.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000374345.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000374345.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000374345.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000374345.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000374345.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000374345.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000374345.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000374345.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000374345.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000422009.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000422009.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000422009.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000422009.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000422009.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000422009.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000422009.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000422009.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000422009.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000422009.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000422009.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000422009.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000424444.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000424444.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000424444.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000432650.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000432650.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000432650.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000432650.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000432650.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000432650.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000432650.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000444723.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000444723.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000444723.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000444723.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000444723.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000444723.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000444723.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000444723.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000444723.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000444723.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000444723.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000450773.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000450773.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000450773.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000450773.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000450773.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | NM_001145350.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | NM_001145350.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | NM_001145350.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | NM_001145350.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | NM_001145350.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | NM_001145350.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | NM_001145350.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | NM_001145350.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | NM_001145350.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | NM_001145350.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | NM_001145350.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | NR_026933.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | NR_026933.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | NR_026933.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | NR_026933.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | NR_026933.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | NR_026933.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | NR_026933.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | NR_026933.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | NR_026933.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | NR_026933.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | NR_026933.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | NR_026933.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | NR_026933.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | NR_026933.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | NR_026933.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | NR_026933.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | NR_026933.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | NR_026933.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | NR_026933.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | NR_026933.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | NR_026933.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | NR_026933.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000162577.hg.1 | ENST00000422009.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000162577.hg.1 | NR_026933.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000162577.hg.1 | TC2000008975.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2000162577.hg.1 | ENST00000422009.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000162577.hg.1 | NR_026933.2 | Refseq |
