Assay Details
| Gene Symbol: SMARCAL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005246632.1 | XP_005246689.1 | 2 - 3 | 260 | 106 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005246632.1 transcripts.
Set Membership: |
|

SMARCAL1
Genomic Map