Assay Details
| Gene Symbol: ATP6V0A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130020.1 | NP_001123492.1 | 17 - 18 | 2196 | 61 | |
| NM_001130021.1 | NP_001123493.1 | 17 - 18 | 2175 | 61 | ||
| NM_005177.3 | NP_005168.2 | 17 - 18 | 2175 | 61 | ||
| XM_005257459.1 | XP_005257516.1 | 17 - 18 | 2178 | 61 | ||
| XM_005257461.1 | XP_005257518.1 | 17 - 18 | 2178 | 61 | ||
| XM_005257463.2 | XP_005257520.1 | 17 - 18 | 2178 | 61 | ||
| XM_017024766.1 | XP_016880255.1 | 17 - 18 | 2178 | 61 | ||
| XM_017024767.1 | XP_016880256.1 | 17 - 18 | 2141 | 61 | ||
| XM_017024768.1 | XP_016880257.1 | 17 - 18 | 2141 | 61 | ||
| XM_017024770.1 | XP_016880259.1 | 17 - 18 | 2141 | 61 | ||
| XM_017024771.1 | XP_016880260.1 | 17 - 18 | 2141 | 61 | ||
| XM_017024772.1 | XP_016880261.1 | 16 - 17 | 1988 | 61 | ||
| XM_017024773.1 | XP_016880262.1 | 16 - 17 | 2569 | 61 | ||
| XM_017024774.1 | XP_016880263.1 | 17 - 18 | 2169 | 61 | ||
| GenBank mRNA | AK223554.1 | - | 17 - 18 | 2142 | 61 | |
| AK294512.1 | - | 16 - 17 | 2002 | 61 | ||
| AK294789.1 | - | 13 - 14 | 1656 | 61 | ||
| AK295682.1 | - | 17 - 18 | 2099 | 61 | ||
| AK299784.1 | - | 16 - 17 | 2014 | 61 | ||
| AK316305.1 | - | 17 - 18 | 2166 | 61 | ||
| BC032398.1 | - | 17 - 18 | 2107 | 5195776 | 61 | |
| BX648978.1 | - | 17 - 18 | 2158 | 61 | ||
| CR627443.1 | - | 10 - 11 | 1083 | 61 | ||
| L78933.1 | - | 17 - 18 | 2177 | 61 | ||
| Z71460.1 | - | 17 - 18 | 2177 | 61 | ||
More Information
Set Membership: |
|

ATP6V0A1
Genomic Map