Assay Details
| Gene Symbol: YAE1D1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282446.1 | NP_001269375.1 | 1 - 2 | 164 | 71 | |
| NM_020192.4 | NP_064577.1 | 1 - 2 | 164 | 71 | ||
| GenBank mRNA | AF226046.1 | - | 1 - 2 | 130 | 71 | |
| AK293511.1 | - | 1 - 2 | 134 | 71 | ||
| AK314937.1 | - | 1 - 2 | 136 | 71 | ||
| BC022043.1 | - | 1 - 2 | 134 | 4734433 | 71 | |
| BM767075.1 | - | 1 - 2 | 164 | 71 | ||
| CR457198.1 | - | 1 - 2 | 121 | 71 | ||
More Information
Set Membership: |
|

YAE1D1
Genomic Map