Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR07023419.hg.1 |
TC07001559.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07023419.hg.1 |
ENST00000265361.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07023419.hg.1 |
ENST00000419255.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07023419.hg.1 |
ENST00000544525.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07023419.hg.1 |
NM_006379.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700182911.hg.1 |
ENST00000265361.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700182911.hg.1 |
ENST00000419255.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700182911.hg.1 |
NM_006379.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700182911.hg.1 |
ENST00000265361.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700182911.hg.1 |
ENST00000419255.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700182911.hg.1 |
NM_006379.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700182911.hg.1 |
TC0700011626.hg.1 |
Clariom D TC |