Assay Details
| Gene Symbol: RPA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001297558.1 | NP_001284487.1 | 2 - 3 | 282 | 71 | |
| NM_002946.4 | NP_002937.1 | 2 - 3 | 475 | 71 | ||
| GenBank mRNA | AK298759.1 | - | 2 - 3 | 253 | 71 | |
| AK312956.1 | - | 2 - 3 | 239 | 71 | ||
| BC001630.1 | - | 2 - 3 | 212 | 3538351 | 71 | |
| BC012157.1 | - | 2 - 3 | 264 | 4563651 | 71 | |
| BC021257.2 | - | 2 - 3 | 403 | 4111194 | 71 | |
| BX333932.2 | - | 2 - 3 | 204 | 71 | ||
| CR450348.1 | - | 1 - 2 | 121 | 71 | ||
| DB025488.1 | - | 2 - 3 | 475 | 71 | ||
| J05249.1 | - | 2 - 3 | 198 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

RPA2
Genomic Map