Assay Details
| Gene Symbol: TRAPPC6A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270891.1 | NP_001257820.1 | 2 - 3 | 176 | 121 | |
| NM_024108.2 | NP_077013.1 | 2 - 3 | 218 | 121 | ||
| XM_017027295.1 | XP_016882784.1 | 2 - 3 | 277 | 121 | ||
| GenBank mRNA | AF161407.1 | - | 2 - 3 | 170 | 121 | |
| BC001907.1 | - | 2 - 3 | 202 | 3534111 | 121 | |
| BC004450.1 | - | 2 - 3 | 203 | 2820830 | 121 | |
| CN256687.1 | - | 2 - 3 | 188 | 121 | ||
| FJ418644.1 | - | 2 - 3 | 158 | 121 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

TRAPPC6A
Genomic Map