Assay Details
Gene Symbol: PAPSS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015880.1 | NP_001015880.1 | 2 - 3 | 410 | 77 | |
NM_004670.3 | NP_004661.2 | 2 - 3 | 410 | 77 | ||
GenBank mRNA | AF074331.1 | - | 2 - 3 | 210 | 77 | |
AF091242.1 | - | 2 - 3 | 227 | 77 | ||
AF150754.2 | - | 2 - 3 | 227 | 77 | ||
AF173365.1 | - | 2 - 3 | 195 | 77 | ||
AF313907.1 | - | 2 - 3 | 147 | 77 | ||
AK309262.1 | - | 2 - 3 | 219 | 77 | ||
AK312556.1 | - | 2 - 3 | 251 | 77 | ||
BC009894.2 | - | 2 - 3 | 258 | 2988345 | 77 | |
BQ221395.1 | - | 2 - 3 | 418 | 77 | ||
CB960618.1 | - | 2 - 3 | 289 | 77 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC10000621.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
sulfate assimilation
skeletal system development
blood coagulation
phosphorylation
3'-phosphoadenosine 5'-phosphosulfate biosynthetic process
bone development
adenylylsulfate kinase activity
sulfate adenylyltransferase (ATP) activity
ATP binding
nucleotidyltransferase activity
sulfate assimilation
skeletal system development
blood coagulation
phosphorylation
3'-phosphoadenosine 5'-phosphosulfate biosynthetic process
bone development
adenylylsulfate kinase activity
sulfate adenylyltransferase (ATP) activity
ATP binding
nucleotidyltransferase activity
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR10006812.hg.1 | ENST00000361175.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10006812.hg.1 | ENST00000371981.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10006812.hg.1 | ENST00000427144.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10006812.hg.1 | ENST00000456849.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10006812.hg.1 | ENST00000482258.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10006812.hg.1 | NM_001015880.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10006812.hg.1 | NM_004670.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10006812.hg.1 | TC10000621.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | ENST00000361175.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | ENST00000361175.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | ENST00000361175.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | ENST00000361175.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | ENST00000361175.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | ENST00000361175.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | ENST00000361175.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | ENST00000361175.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | ENST00000361175.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | ENST00000361175.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | ENST00000361175.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | ENST00000361175.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | ENST00000361175.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | ENST00000361175.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | ENST00000361175.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | ENST00000361175.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | ENST00000361175.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | ENST00000361175.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | ENST00000456849.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | ENST00000456849.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | ENST00000456849.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | ENST00000456849.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | ENST00000456849.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | ENST00000456849.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | ENST00000456849.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | ENST00000456849.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | ENST00000456849.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | ENST00000456849.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | ENST00000456849.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | ENST00000456849.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | ENST00000456849.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | ENST00000456849.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | ENST00000456849.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | ENST00000456849.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | ENST00000456849.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | ENST00000465996.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | ENST00000465996.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | ENST00000465996.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | ENST00000482258.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | ENST00000482258.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | ENST00000482258.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | ENST00000482258.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | NM_001015880.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | NM_001015880.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | NM_001015880.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | NM_001015880.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | NM_001015880.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | NM_001015880.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | NM_001015880.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | NM_001015880.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | NM_001015880.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | NM_001015880.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | NM_001015880.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | NM_001015880.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | NM_001015880.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | NM_001015880.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | NM_001015880.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | NM_001015880.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | NM_001015880.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | NM_004670.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | NM_004670.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | NM_004670.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | NM_004670.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | NM_004670.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | NM_004670.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | NM_004670.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | NM_004670.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | NM_004670.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | NM_004670.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | NM_004670.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | NM_004670.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | NM_004670.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | NM_004670.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | NM_004670.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | NM_004670.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | NM_004670.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10006811.hg.1 | ENST00000361175.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10006811.hg.1 | ENST00000371981.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10006811.hg.1 | ENST00000427144.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10006811.hg.1 | ENST00000456849.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10006811.hg.1 | ENST00000482258.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10006811.hg.1 | NM_001015880.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10006811.hg.1 | NM_004670.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10006811.hg.1 | TC10000621.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | ENST00000361175.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | ENST00000361175.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | ENST00000361175.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | ENST00000361175.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | ENST00000361175.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | ENST00000361175.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | ENST00000361175.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | ENST00000361175.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | ENST00000361175.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | ENST00000361175.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | ENST00000361175.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | ENST00000361175.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | ENST00000361175.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | ENST00000361175.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | ENST00000361175.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | ENST00000361175.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | ENST00000361175.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | ENST00000361175.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | ENST00000456849.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | ENST00000456849.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | ENST00000456849.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | ENST00000456849.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | ENST00000456849.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | ENST00000456849.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | ENST00000456849.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | ENST00000456849.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | ENST00000456849.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | ENST00000456849.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | ENST00000456849.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | ENST00000456849.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | ENST00000456849.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | ENST00000456849.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | ENST00000456849.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | ENST00000456849.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | ENST00000456849.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | ENST00000465996.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | ENST00000465996.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | ENST00000465996.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | ENST00000482258.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | ENST00000482258.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | ENST00000482258.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | ENST00000482258.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | NM_001015880.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | NM_001015880.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | NM_001015880.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | NM_001015880.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | NM_001015880.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | NM_001015880.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | NM_001015880.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | NM_001015880.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | NM_001015880.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | NM_001015880.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | NM_001015880.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | NM_001015880.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | NM_001015880.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | NM_001015880.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | NM_001015880.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | NM_001015880.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | NM_001015880.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | NM_004670.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | NM_004670.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | NM_004670.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | NM_004670.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | NM_004670.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | NM_004670.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | NM_004670.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | NM_004670.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | NM_004670.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | NM_004670.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | NM_004670.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | NM_004670.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | NM_004670.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | NM_004670.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | NM_004670.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | NM_004670.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | NM_004670.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000156590.hg.1 | ENST00000361175.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000156590.hg.1 | ENST00000456849.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000156590.hg.1 | ENST00000465996.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000156590.hg.1 | ENST00000482258.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000156590.hg.1 | NM_001015880.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000156590.hg.1 | NM_004670.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000156590.hg.1 | ENST00000361175.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000156590.hg.1 | ENST00000456849.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000156590.hg.1 | ENST00000465996.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000156590.hg.1 | ENST00000482258.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000156590.hg.1 | NM_001015880.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000156590.hg.1 | NM_004670.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000156590.hg.1 | TC1000008354.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1000156591.hg.1 | ENST00000361175.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000156591.hg.1 | ENST00000456849.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000156591.hg.1 | ENST00000482258.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000156591.hg.1 | NM_001015880.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000156591.hg.1 | NM_004670.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000156591.hg.1 | ENST00000361175.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000156591.hg.1 | ENST00000456849.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000156591.hg.1 | ENST00000482258.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000156591.hg.1 | NM_001015880.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000156591.hg.1 | NM_004670.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000156591.hg.1 | TC1000008354.hg.1 | Clariom D TC |
